Merck
  • The effect of non-enzymatic glycation on the unfolding of human serum albumin.
The effect of non-enzymatic glycation on the unfolding of human serum albumin.
Archives of biochemistry and biophysics (2005-11-29)
Deanna L Mendez, Russell A Jensen, Laura A McElroy, Jose M Pena, Raymond M Esquerra
ABSTRACT
We monitored the unfolding of human serum albumin (HSA) and glycated human serum albumin (gHSA) subjected to guanidine hydrochloride (GndHCl) by using fluorescence and circular dichroism (CD) spectroscopy. A two-state model with sloping baselines best described the Trp-214 fluorescence unfolding measurements, while a three-state model best described the far-UV CD unfolding data. Glycation of HSA increased the [D](50%) point by approximately 0.20M. This corresponded to an increase in the free energy of unfolding of gHSA relative to HSA of 2.6kJ/mol. The intrinsic fluorescence of Trp-214 in gHSA is 0.72 of that of HSA and the far-UV CD spectrum of gHSA is nearly identical to that of HSA. These results showed that glycation altered the local structure around Trp-214 while not significantly impacting the secondary structure, and this alteration translated into an overall change in the stability of gHSA compared to HSA.
Sigma-Aldrich
m-Aminophenylboronic acid–Agarose, aqueous suspension
Sigma-Aldrich
Albumin from human serum, lyophilized powder, Fatty acid free, Globulin free, ≥99% (agarose gel electrophoresis)