Merck
  • Structure analysis and antimutagenic activity of a novel salt-soluble polysaccharide from Auricularia polytricha.
Structure analysis and antimutagenic activity of a novel salt-soluble polysaccharide from Auricularia polytricha.
Journal of the science of food and agriculture (2013-04-05)
Jie Zhou, Yun Chen, Mengting Xin, Qinyao Luo, Jialu Gu, Mengtian Zhao, Xiaolan Xu, Xuanqi Lu, Guanglei Song
ABSTRACT
Auricularia polytricha is known to be a highly nutritious foodstuff. We report here the purification, structure characterization and antimutagenic activity in vivo of a 0.9% NaCl solution-soluble polysaccharide (SSP) from the mycelia of A. polytricha. Analysis by high-performance liquid chromatography coupled to a TSK-G5000PWXL column and gel filtration chromatography on Sephacryl S-400 HR indicated that SSP is homogeneous with an average molecular weight of about 9.30 × 10(5) Da. The structure of SSP was revealed by chemical methods, Fourier transform infrared spectroscopy and nuclear magnetic resonance spectroscopy. Results indicated that SSP is a glucan consisting of a1,3-β-glucan, 1,6-α-glucan, 1,4-α-glucan and 1,3-α-glucan backbone with a single 1,6-α-d-glucopyranosyl side-branching unit on every nine residues, on average, along the main chain. Atomic force microscopy indicates the presence of macromolecular species in morphology and shows a clear association of prolate particle. Meanwhile, SSP was found to significantly preventing micronuclei in polychromatic erythrocytes and reticulocytes of mice (P < 0.01). These results indicate that polysaccharide SSP from A. polytricha exhibits antimutagenic activity against the in vivo DNA-damaging effect of the indirectly acting alkylating agent cyclophosphamide.
Sigma-Aldrich
Periodic acid, 99.999% trace metals basis
Sigma-Aldrich
Periodic acid, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Periodic acid, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Periodic acid, puriss. p.a., ACS reagent, crystallized, ≥99.0% (RT)
Sigma-Aldrich
Sodium periodate, ACS reagent, ≥99.8%
Sigma-Aldrich
Periodic acid, ACS reagent, 99%
Sigma-Aldrich
Sodium (meta)periodate, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Sodium (meta)periodate, ≥99.0%