Merck
  • Applicability of Lactobacillus plantarum IMDO 788 as a starter culture to control vegetable fermentations.
Applicability of Lactobacillus plantarum IMDO 788 as a starter culture to control vegetable fermentations.
Journal of the science of food and agriculture (2013-04-23)
Dorrit Wouters, Silvia Grosu-Tudor, Medana Zamfir, Luc De Vuyst
ABSTRACT
Fermentation of vegetables and fruits is a traditional preservation technique, e.g. in Eastern Europe. Although usually spontaneous fermentation processes are applied, the addition of lactic acid bacteria (LAB) starter cultures could accelerate processing and improve the consistency and quality of the end-products. The application of Lactobacillus plantarum IMDO 788 as a starter culture strain for cauliflower and mixed vegetable fermentations resulted in accelerated acidification as compared with the spontaneous fermentations. The strain dominated the background microbiota throughout the process, whereas the spontaneous fermentations were characterised by widely variable species diversity. During the spontaneous fermentations, almost all carbohydrates were converted into lactic acid, ethanol, mannitol and acetic acid, indicating the participation of both heterofermentative and homofermentative LAB species. During the starter culture-added fermentations, residual carbohydrates were found and lactic acid and ethanol were the main end-metabolites. Vegetable-associated aromas, ethyl acetate and isoamyl acetate were produced during all fermentations. The high concentration of ethanol and the production of ethyl acetate and isoamyl acetate suggested the involvement of yeasts during all fermentations. Lactobacillus plantarum IMDO 788 was an adequate starter culture strain for vegetable fermentations, prevailing over endogenous LAB communities. Further optimisation of the starter culture formulation is necessary to avoid yeast growth.
Sigma-Aldrich
Isopentyl acetate, anhydrous, ≥99%
Supelco
Isopentyl acetate, analytical standard
Sigma-Aldrich
Isopentyl acetate, mixture of isomers, reagent grade, 98%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Isoamyl acetate, ≥95%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Isoamyl acetate, natural, ≥97%, FCC, FG
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%