Merck
  • Alcohol plus chlorhexidine is more efficient than alcohol alone for phenol-based chemical matricectomy: an in vitro study.
Alcohol plus chlorhexidine is more efficient than alcohol alone for phenol-based chemical matricectomy: an in vitro study.
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.] (2013-05-04)
Damian Cordoba Diaz, Ricardo Becerro de Bengoa Vallejo, Marta Elena Losa Iglesias, Manuel Cordoba Diaz
ABSTRACT
A phenolization approach is often chosen for treatment of ingrown toenails. Many reports describe lavage of the wound with alcohol to neutralize any residual phenol. The aim of our study was to compare the suitability and effectiveness of a solution consisting of alcohol plus chlorhexidine with that of alcohol alone in a lavage step during the treatment of ingrown toenails using a phenolization procedure. We performed an in vitro study using human matrix and a diffusion cell apparatus to measure the amount of phenol remaining after various lavage washes. The effect of phenol evaporation was also examined. When phenol was irrigated with alcohol alone, total phenol recovered was 29.9 ± 4.8 mg (62.9% of the initial amount). When the phenol was irrigated with isopropyl alcohol plus 0.5% chlorhexidine, 39.8 ± 1.7 mg (83.8%) of phenol was recovered. A lavage step including alcohol and chlorhexidine is more effective than alcohol alone for removal of excess phenol. The addition of chlorhexidine also contributed to inhibition of the action of phenol. This approach provides a suitable and effective means of diluting and removing any excess or residual phenol from the exposed area.
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Chlorhexidine dihydrochloride, ≥98%
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Chlorhexidine dihydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Chlorhexidine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%