Merck
  • Physicochemical and biological evaluation of poly(ethylene glycol) methacrylate grafted onto poly(dimethyl siloxane) surfaces for prosthetic devices.
Physicochemical and biological evaluation of poly(ethylene glycol) methacrylate grafted onto poly(dimethyl siloxane) surfaces for prosthetic devices.
Colloids and surfaces. B, Biointerfaces (2013-05-11)
Sara Gonçalves, Ana Leirós, Theo van Kooten, Fernando Dourado, Lígia R Rodrigues
ABSTRACT
Poly(dimethyl siloxane) (PDMS) was surface-polymerized with poly(ethylene glycol)methacrylate (PEGMA) by surface-initiated atom transfer radical polymerization (SI-ATRP) in aqueous media at room temperature. Modification of the PDMS surface followed a three-step procedure: (i) PDMS surface hydroxylation by UV/ozone exposure, immediately followed by (ii) covalent attachment of the initiator, 1-trichlorosilyl-2-(chloromethylphenyl)ethane, onto the hydroxylated PDMS, via chemical vapor deposition; finally (iii) PDMS surface-polymerization of PEGMA by ATRP. Modified PDMS was characterized by water contact angle measurement, SEM, FTIR-ATR, and XPS. Results showed that modified surfaces had a hydrophilic character, given the water contact angles around 60°; FTIR-ATR and XPS analysis confirmed the presence of polymerized PEGMA on the surface of PDMS and the adhesion of Staphylococcus aureus GB 2/1 and Streptococcus salivarius GB 24/9 onto the modified surfaces was inhibited 94% and 81%, respectively. Finally, the modified PDMS showed no evidence of cytotoxic effects in in vitro assays using human skin fibroblasts.
Product Description
Silicone oil, for melting point and boiling point apparatuses
Sigma-Aldrich
Silicone oil, viscosity 350 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 10 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 1,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 500 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 10,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 100,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 100 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 60,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 50 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 20 cSt (25 °C)
Sigma-Aldrich
Silicone oil
Sigma-Aldrich
Silicone oil, viscosity 5 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 30,000 cSt (25 °C)