Merck
  • A sustainable two-phase procedure for V-catalyzed toluene oxidative bromination with H2O2-KBr.
A sustainable two-phase procedure for V-catalyzed toluene oxidative bromination with H2O2-KBr.
Dalton transactions (Cambridge, England : 2003) (2013-06-12)
Pierluca Galloni, Marco Mancini, Barbara Floris, Valeria Conte
ABSTRACT
A sustainable V(V) and Mo(VI) catalysed two-phase procedure for bromination of toluene under quite mild conditions is proposed; H2O2 is the primary oxidant and KBr is the bromine source; metal precursors are commercially available salts. The reaction is efficient without any additional solvent. By using PhCH3 as a solvent/substrate good yields, together with interesting selectivity toward the formation of PhCH2Br, are obtained with both metal ions. Recycling of the catalytic phase is also possible. Useful information on the V-peroxido chemistry was obtained.
Product Description
Supelco
Toluene solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Vanadium, powder, −325 mesh, 99.5% trace metals basis
Sigma-Aldrich
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis
Supelco
Toluene, analytical standard
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%