Merck
  • Nicotinamide supplementation induces detrimental metabolic and epigenetic changes in developing rats.
Nicotinamide supplementation induces detrimental metabolic and epigenetic changes in developing rats.
The British journal of nutrition (2013-06-19)
Da Li, Yan-Jie Tian, Jing Guo, Wu-Ping Sun, Yong-Zhi Lun, Ming Guo, Ning Luo, Yu Cao, Ji-Min Cao, Xiao-Jie Gong, Shi-Sheng Zhou
ABSTRACT
Ecological evidence suggests that niacin (nicotinamide and nicotinic acid) fortification may be involved in the increased prevalence of obesity and type 2 diabetes, both of which are associated with insulin resistance and epigenetic changes. The purpose of the present study was to investigate nicotinamide-induced metabolic changes and their relationship with possible epigenetic changes. Male rats (5 weeks old) were fed with a basal diet (control group) or diets supplemented with 1 or 4 g/kg of nicotinamide for 8 weeks. Low-dose nicotinamide exposure increased weight gain, but high-dose one did not. The nicotinamide-treated rats had higher hepatic and renal levels of 8-hydroxy-2'-deoxyguanosine, a marker of DNA damage, and impaired glucose tolerance and insulin sensitivity when compared with the control rats. Nicotinamide supplementation increased the plasma levels of nicotinamide, N1-methylnicotinamide and choline and decreased the levels of betaine, which is associated with a decrease in global hepatic DNA methylation and uracil content in DNA. Nicotinamide had gene-specific effects on the methylation of CpG sites within the promoters and the expression of hepatic genes tested that are responsible for methyl transfer reactions (nicotinamide N-methyltransferase and DNA methyltransferase 1), for homocysteine metabolism (betaine-homocysteine S-methyltransferase, methionine synthase and cystathionine β-synthase) and for oxidative defence (catalase and tumour protein p53). It is concluded that nicotinamide-induced oxidative tissue injury, insulin resistance and disturbed methyl metabolism can lead to epigenetic changes. The present study suggests that long-term high nicotinamide intake (e.g. induced by niacin fortification) may be a risk factor for methylation- and insulin resistance-related metabolic abnormalities.
Sigma-Aldrich
Nicotinic acid, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinic acid, meets USP testing specifications
Sigma-Aldrich
Nicotinic acid, ≥98%
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent
Sigma-Aldrich
Nicotinic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Supelco
Niacin (Nicotinic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Nicotinic acid, analytical standard
Sigma-Aldrich
Choline chloride, ≥98%
Sigma-Aldrich
Choline chloride, ≥99%
Sigma-Aldrich
Betaine, ≥98% (perchloric acid titration)
Sigma-Aldrich
Betaine hydrochloride, ≥99%
Sigma-Aldrich
Choline chloride, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Choline chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Supelco
Choline chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
1-Methylnicotinamide chloride, analytical standard
Sigma-Aldrich
Betaine, BioUltra, ≥99.0% (NT)