Merck
  • Photoinduced charge transfer process in p-Cu2O/n-Cu2O homojunction film and its photoelectric gas-sensing properties.
Photoinduced charge transfer process in p-Cu2O/n-Cu2O homojunction film and its photoelectric gas-sensing properties.
Journal of colloid and interface science (2013-06-20)
Tengfei Jiang, Tengfeng Xie, Wanshi Yang, Haimei Fan, Dejun Wang
ABSTRACT
In this study, the p-Cu2O/n-Cu2O homojunction film was prepared by a simple two-step electrodeposition method. The photoinduced charge transfer process in p-Cu2O/n-Cu2O homojunction film was studied using surface photovoltage technique. Then, the p-Cu2O/n-Cu2O homojunction film was assembled into a photoelectric gas sensor for sensing acetaldehyde both under 405 nm and 532 nm light irradiation at room temperature. For 532 nm light irradiation, excess photoinduced electrons migrate to the irradiated n-Cu2O side affected by the interfacial built-in electric field and promote desorption of acetaldehyde, which results in a higher sensitivity than that under 405 nm light. Our results demonstrated that constructing junction structure in photoelectric gas-sensing materials is an effective approach to achieve high sensitivity at room temperature.
Sigma-Aldrich
Acetaldehyde, natural, FG
Sigma-Aldrich
Acetaldehyde, puriss. p.a., anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Acetaldehyde, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Acetaldehyde, ACS reagent, ≥99.5%
Supelco
Acetaldehyde, PESTANAL®, analytical standard
Sigma-Aldrich
Copper(II) oxide, 99.999% trace metals basis
Sigma-Aldrich
Acetaldehyde solution, natural, 50 wt. % ethanol, FG
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Copper(II) oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Copper(II) oxide, powder, 99.995% trace metals basis
Sigma-Aldrich
Copper(II) oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF
Sigma-Aldrich
Copper(II) oxide, needles, mixture of CuO and Cu2O, ACS reagent
Sigma-Aldrich
Copper(II) oxide, ACS reagent, ≥99.0%
Sigma-Aldrich
Copper(II) oxide on alumina, 14-30 mesh, extent of labeling: 13 wt. % loading
Sigma-Aldrich
Copper(II) oxide, powder, <10 μm, 98%