Merck
  • Subchronic exposure to ethyl tertiary butyl ether resulting in genetic damage in Aldh2 knockout mice.
Subchronic exposure to ethyl tertiary butyl ether resulting in genetic damage in Aldh2 knockout mice.
Toxicology (2013-07-03)
Zuquan Weng, Megumi Suda, Katsumi Ohtani, Nan Mei, Toshihiro Kawamoto, Tamie Nakajima, Rui-Sheng Wang
ABSTRACT
Ethyl tertiary butyl ether (ETBE) is biofuel additive recently used in Japan and some other countries. Limited evidence shows that ETBE has low toxicity. Acetaldehyde (AA), however, as one primary metabolite of ETBE, is clearly genotoxic and has been considered to be a potential carcinogen. The aim of this study was to evaluate the effects of ALDH2 gene on ETBE-induced genotoxicity and metabolism of its metabolites after inhalation exposure to ETBE. A group of wild-type (WT) and Aldh2 knockout (KO) C57BL/6 mice were exposed to 500ppm ETBE for 1-6h, and the blood concentrations of ETBE metabolites, including AA, tert-butyl alcohol and 2-methyl-1,2-propanediol, were measured. Another group of mice of WT and KO were exposed to 0, 500, 1750, or 5000ppm ETBE for 6h/day with 5 days per weeks for 13 weeks. Genotoxic effects of ETBE in these mice were measured by the alkaline comet assay, 8-hydroxyguanine DNA-glycosylase modified comet assay and micronucleus test. With short-term exposure to ETBE, the blood concentrations of all the three metabolites in KO mice were significantly higher than the corresponding concentrations of those in WT mice of both sexes. After subchronic exposure to ETBE, there was significant increase in DNA damage in a dose-dependent manner in KO male mice, while only 5000ppm exposure significantly increased DNA damage in male WT mice. Overall, there was a significant sex difference in genetic damage in both genetic types of mice. These results showed that ALDH2 is involved in the detoxification of ETBE and lack of enzyme activity may greatly increase the sensitivity to the genotoxic effects of ETBE, and male mice were more sensitive than females.
Sigma-Aldrich
tert-Butanol, puriss. p.a., ACS reagent, ≥99.7% (GC)
Sigma-Aldrich
Acetaldehyde, natural, FG
Supelco
Acetaldehyde, PESTANAL®, analytical standard
Sigma-Aldrich
tert-Butanol, anhydrous, ≥99.5%
Sigma-Aldrich
Acetaldehyde, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetaldehyde, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Acetaldehyde, puriss. p.a., anhydrous, ≥99.5% (GC)
Supelco
tert-Butanol, analytical standard
Sigma-Aldrich
tert-Butanol, suitable for HPLC, ≥99.5%
Sigma-Aldrich
tert-Butanol, ACS reagent, ≥99.0%
Sigma-Aldrich
tert-Butanol, TEBOL® 99, ≥99.3%
Sigma-Aldrich
tert-Butanol, ≥99% (GC)
Sigma-Aldrich
Acetaldehyde solution, natural, 50 wt. % ethanol, FG
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Supelco
tert-Butyl ethyl ether, analytical standard
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
tert-Butyl ethyl ether, purum, ≥97.0% (GC)
Sigma-Aldrich
tert-Butyl ethyl ether, 99%
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF