Merck
  • Hydrotalcite composites for an effective fluoride buccal administration: a new technological approach.
Hydrotalcite composites for an effective fluoride buccal administration: a new technological approach.
International journal of pharmaceutics (2013-07-16)
Luana Perioli, Morena Nocchetti, Paola Giannelli, Cinzia Pagano, Maria Bastianini
ABSTRACT
The aim of this work was to develop new mucoadhesive buccal patches containing an inorganic fluorinated compound, MgAl-F, intended for decay prevention. Firstly MgAl-F was synthesized and characterized, then the patches were prepared starting from a physical blend of mucoadhesive polymers (NaCMC and polycarbophil) in which MgAl-F was dispersed in different amounts in order to obtain the films. The prepared mucoadhesive patches were characterized in terms of swelling capacity, mucoadhesion force and time, surface morphology and in vitro release studies. Moreover, the organoleptic properties and acceptability have been evaluated by in vivo application. The performed studies demonstrated that the proposed formulations are practical, manageable, flexible and adaptable to the biological substrate showing, at the same time, good organoleptic properties. Moreover, the presence of the MgAl-F is able to decrease the strong adhesion of the employed polymers, reducing pain and irritations resulting in a high patient acceptability. Data obtained from release studies revealed that the application of small patch portions is enough able to release, for a prolonged time, an amount of fluoride ions able to reach the efficacious dose. These observations suggest the applicability of such formulations for buccal administration of different active ingredients.
Sigma-Aldrich
Magnesium hydroxide, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Carboxymethylcellulose sodium, meets USP testing specifications, Medium viscosity
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 1.2
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 0.7
Sigma-Aldrich
Sodium carboxymethyl cellulose, viscosity 2500-6000 cP 
Sigma-Aldrich
Sodium carboxymethyl cellulose, viscosity 50-200 cP , c=4% H2O at 25­°C
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 0.9
Sigma-Aldrich
Magnesium hydroxide, reagent grade, 95%
Sigma-Aldrich
Aluminum hydroxide, reagent grade
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Carboxymethylcellulose sodium salt, low viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, Medium viscosity
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Supelco
Fluoride ion solution for ISE, 0.1 M F-, for ion-selective electrodes
Sigma-Aldrich
Carboxymethylcellulose sodium salt, medium viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, ultra high viscosity, highly purified
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Carboxymethylcellulose sodium salt, High viscosity