Merck
  • Synthesis and characterisation of galactosyl glycerol by β-galactosidase catalysed reverse hydrolysis of galactose and glycerol.
Synthesis and characterisation of galactosyl glycerol by β-galactosidase catalysed reverse hydrolysis of galactose and glycerol.
Food chemistry (2013-07-23)
Wei Wei, Danping Qi, Hai-zhen Zhao, Zhao-xin Lu, Fengxia Lv, Xiaomei Bie
ABSTRACT
A study of an enzymatic method for the production of galactosylglycerol is described. The effects of enzyme sources, enzyme amount, reaction temperature, reaction time, substrate ratio of glycerol to galactose, buffer content and pH on galactosylglycerol yield (mg/ml) were investigated. Under the optimum reaction conditions of β-galactosidases from Kluyveromyces lactis 240 U/ml, temperature 40 °C, time 24 h, buffer amount 60% (percent volume of buffer to that of substrates, pH 6.5), and the substrate molar ratio of 10 (glycerol16 mmol:galactose 1.6 mmol), the yield of galactosylglycerol was up to 116.47 mg/ml (galactose conversion 55.88%). The product was purified by activated charcoal and Sephadex G-15 column chromatography, up to 96%. The purified galactosylglycerol was fully characterised by MS and NMR, and identified as a mixture of (2R)- and (2S)- 3-O-β-D-galactopyranosyl-glycerol.
Product Description
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)