Merck
Dermatopontin regulates fibrin formation and its biological activity.
The Journal of investigative dermatology (2013-07-24)
Weimin Wu, Osamu Okamoto, Aiko Kato, Noritaka Matsuo, Motoyoshi Nomizu, Hidekatsu Yoshioka, Sakuhei Fujiwara
ABSTRACT
Dermatopontin (DP) is a small extracellular matrix component in the dermis. Fibrin is a major component of a provisional matrix that is formed just after wounding. Previously, we found that DP was present in the provisional matrix, and it interacted with fibrin. Here, we examined the role of DP on fibrin function. DP interacted with both the fibrin monomer and fibrils, and was incorporated into the fibrils during fibrin formation. A DP sequence, PHGQVVVAVRS, was identified as a fibrin-binding site, and a globular D domain of fibrin was the binding site for DP. DP accelerated fibrin fibril formation into structurally modified fibrils. Fibrin fibrils formed in the presence of DP enhanced both endothelial cell attachment and cell spreading. The attached cells developed a more organized cytoskeleton when compared with those that attached to fibrin fibrils only. The main receptor for cell adhesion was identified as αvβ3 integrin, and a cooperating receptor was a β1-containing integrin species, probably α5β1 integrin. These results indicate that DP can modify certain biological functions of fibrin, and thus a another function of this extracellular matrix protein was revealed. In addition, the fibrin-DP complex might become useful for developing an improved artificial matrix for improving wound healing.
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone 6S6, clone 6S6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin αVβ3 Antibody, clone LM609, clone LM609, Chemicon®, from mouse