Merck
  • Sclerosant foam structure and stability is strongly influenced by liquid air fraction.
Sclerosant foam structure and stability is strongly influenced by liquid air fraction.
European journal of vascular and endovascular surgery : the official journal of the European Society for Vascular Surgery (2013-09-03)
E Cameron, T Chen, D E Connor, M Behnia, K Parsi
ABSTRACT
To determine the effects of sclerosant foam preparation and composition on foam structure, the time course of liquid drainage, and foam coarsening. Sodium tetradecyl sulphate (STS) and polidocanol (POL) foams were investigated in a range of concentrations (0.5-3%) and liquid-plus-air fractions (LAF; 1 + 2 to 1 + 8). Foam was injected into a vein simulation model (polyvinyl chloride tubing, inner diameter 3 mm, constant pressure 5-7 mmHg) filled with saline or blood. Liquid drainage, bubble count, and diameter were measured and documented by serial photography. Liquid drainage was faster in the vertical position than the horizontal one. In all variations, very small bubbles (diameter <30 μm) were generated initially that coarsened to form micro-foams (<250 μm). By 3 minutes mini-foams (>250 μm) and by 7.5 minutes macro-foams (>500 μm) were formed. Following injection, the upper regions of foam coarsened faster as liquid drained to the bottom of the vessel. Wet preparations produced significantly smaller bubbles. Low concentration POL foam produced significantly higher bubble counts and coarsened slower than STS. Foam structure is strongly influenced by the LAF. Despite the initial formation of micro-bubbles in the syringe, mini- and macro-bubbles are formed in target vessels with time post-injection.
Sigma-Aldrich
Poly(vinyl chloride), average Mw ~233,000, average Mn ~99,000
Sigma-Aldrich
Poly(vinyl chloride), average Mw ~62,000, average Mn ~35,000
Sigma-Aldrich
Poly(vinyl chloride), average Mw ~43,000, average Mn ~22,000
Sigma-Aldrich
Poly(vinyl chloride), low molecular weight
Sigma-Aldrich
Poly(vinyl chloride), high molecular weight
Sigma-Aldrich
Brij® L23, main component: tricosaethylene glycol dodecyl ether
Supelco
Poly(vinyl chloride), Selectophore, high molecular weight
Sigma-Aldrich
Brij® L23, suitable for Stein-Moore chromatography
Sigma-Aldrich
Brij® L4, average Mn ~362
Supelco
Poly(vinyl chloride), analytical standard, average Mw 85,000 (Typical), average Mn 40,000 (Typical)
Poly(vinyl chloride), European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
ECO Brij® L23
Sigma-Aldrich
ECO BRIJ® L4, average Mn ~362
Sigma-Aldrich
Thesit®
Sigma-Aldrich
Decaethylene glycol mono­dodecyl ether, nonionic surfactant
Sigma-Aldrich
Tetradecyl sulfate sodium salt, 95%
Sigma-Aldrich
Brij® L23 solution, 30 % (w/v) in H2O