Merck
  • Clinical outcomes of vitamin D deficiency and supplementation in cancer patients.
Clinical outcomes of vitamin D deficiency and supplementation in cancer patients.
Nutrition reviews (2013-09-17)
Laisa Teleni, Jacqueline Baker, Bogda Koczwara, Michael G Kimlin, Euan Walpole, Kathy Tsai, Elizabeth A Isenring
ABSTRACT
Results of recent studies suggest that circulating levels of vitamin D may play an important role in cancer-specific outcomes. The present systematic review was undertaken to determine the prevalence of vitamin D deficiency (<25 nmol/L) and insufficiency (25-50 nmol/L) in cancer patients and to evaluate the association between circulating calcidiol (the indicator of vitamin D status) and clinical outcomes. A systematic search of original, peer-reviewed studies on calcidiol at cancer diagnosis, and throughout treatment and survival, was conducted yielding 4,706 studies. A total of 37 studies met the inclusion criteria for this review. Reported mean blood calcidiol levels ranged from 24.7 to 87.4 nmol/L, with up to 31% of patients identified as deficient and 67% as insufficient. The efficacy of cholecalciferol supplementation for raising the concentration of circulating calcidiol is unclear; standard supplement regimens of <1,000 IU D₃ /day may not be sufficient to maintain adequate concentrations or prevent decreasing calcidiol. Dose-response studies linking vitamin D status to musculoskeletal and survival outcomes in cancer patients are lacking.
Product Description
Supelco
Cholecalciferol (D3), analytical standard
Cholecalciferol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cholecalciferol, meets USP testing specifications
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
Cholecalciferol, analytical standard
Sigma-Aldrich
3-epi-25-Hydroxyvitamin D3, 98% (CP)
Sigma-Aldrich
25-Hydroxycholecalciferol, ≥98% (HPLC)
Supelco
Cholecalciferol (Vitamin D3), Pharmaceutical Secondary Standard; Certified Reference Material