Merck
  • RP-HPLC analytical method development and optimization for quantification of donepezil hydrochloride in orally disintegrating tablet.
RP-HPLC analytical method development and optimization for quantification of donepezil hydrochloride in orally disintegrating tablet.
Pakistan journal of pharmaceutical sciences (2013-09-17)
Kai Bin Liew, Kok Khiang Peh, Yvonne Tze Fung Tan
ABSTRACT
An easy, fast and validated RV-HPLC method was invented to quantify donepezil hydrochloride in drug solution and orally disintegrating tablet. The separation was carried out using reversed phase C-18 column (Agilent Eclipse Plus C-18) with UV detection at 268 nm. Method optimization was tested using various composition of organic solvent. The mobile phase comprised of phosphate buffer (0.01M), methanol and acetonitrile (50:30:20, v/v) adjusted to pH 2.7 with phosphoric acid (80%) was found as the optimum mobile phase. The method showed intraday precision and accuracy in the range of 0.24% to -1.83% and -1.83% to 1.99% respectively, while interday precision and accuracy ranged between 1.41% to 1.81% and 0.11% to 1.90% respectively. The standard calibration curve was linear from 0.125 μg/mL to 16 μg/mL, with correlation coefficient of 0.9997±0.00016. The drug solution was stable under room temperature at least for 6 hours. System suitability studies were done. The average plate count was > 2000, tailing factor <1, and capacity factor of 3.30. The retention time was 5.6 min. The HPLC method was used to assay donepezil hydrochloride in tablet and dissolution study of in-house manufactured donepezil orally disintegrating tablet and original Aricept.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O