Merck
  • Effects of selenium on development, survival, and accumulation in the honeybee (Apis mellifera L.).
Effects of selenium on development, survival, and accumulation in the honeybee (Apis mellifera L.).
Environmental toxicology and chemistry (2013-10-12)
Kristen R Hladun, Osman Kaftanoglu, David R Parker, Khoa D Tran, John T Trumble
ABSTRACT
Apis mellifera L. (Hymenoptera: Apidae) is an important agricultural pollinator in the United States and throughout the world. In areas of selenium (Se) contamination, honeybees may be at risk because of the biotransfer of Se from plant products such as nectar and pollen. Several forms of Se can occur in accumulating plants. In the present study, the toxicity of 4 compounds (selenate, selenite, methylselenocysteine, and selenocystine) to honeybee adult foragers and larvae was assessed using dose-response bioassays. Inorganic forms were more toxic than organic forms for both larvae (lethal concentration [LC50] selenate = 0.72 mg L(-1) , LC50 selenite = 1.0 mg L(-1) , LC50 methylselenocysteine = 4.7 mg L(-1) , LC50 selenocystine = 4.4 mg L(-1) ) and foragers (LC50 selenate = 58 mg L(-1) , LC50 selenite = 58 mg L(-1) , LC50 methylselenocysteine = 161 mg L(-1) , LC50 selenocystine = 148 mg L(-1) ). Inorganic forms of Se caused rapid mortality, and organic forms had sublethal effects on development. Larvae accumulated substantial amounts of Se only at the highest doses, whereas foragers accumulated large quantities at all doses. The present study documented very low larval LC50 values for Se; even modest transfer to brood will likely cause increased development times and mortality. The toxicities of the various forms of Se to honeybee larvae and foragers are discussed in comparison with other insect herbivores and detritivores.
Product Description
Supelco
L-Cystine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Cystine, Wacker Chemie AG, ≥98.5%
SAFC
L-Cystine
Sigma-Aldrich
L-Cystine, ≥99.7% (TLC)
Sigma-Aldrich
L-Cystine, ≥98% (TLC), crystalline
Sigma-Aldrich
L-Cystine, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-101.0%
Cystine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Seleno-L-cystine, 95%
Sigma-Aldrich
Selenous acid, 99.999% trace metals basis
Sigma-Aldrich
Selenous acid, 98%
Sigma-Aldrich
Seleno-DL-cystine, powder
Sigma-Aldrich
Sodium selenate, BioXtra
Sigma-Aldrich
Sodium selenate, purum p.a., ≥98.0% (T)
Sigma-Aldrich
Sodium selenate, ≥95% (elemental analysis)