Merck
  • Evaluation and enhancement of physical stability of semi-solid dispersions containing piroxicam into hard gelatin capsules.
Evaluation and enhancement of physical stability of semi-solid dispersions containing piroxicam into hard gelatin capsules.
Acta poloniae pharmaceutica (2013-10-24)
Ayşegül Karataş, Serife Bekmezci
ABSTRACT
The aim of the study was to investigate the physical stability of the semi-solid dispersions into the hard gelatine capsules prepared with Gelucire 44/14, Labrasol and different additives such as microcrystalline cellulose (MCC), mannitol and lactose (alpha-monohydrate) used for enhancing the stability of the formulations. The master dispersion prepared with only Gelucire 44/14 (20% w/w) and Labrasol (80% w/w) was stored in a refrigerator (5 +/- 3 degrees C), while the modified dispersions with the additives (2% w/w) were kept in a climatic chamber (25 +/- 2 degrees C / 60 +/- 5% RH) for 12 months. Dissolution tests of the semi-solid dispersions were performed in media with different pH's immediatly after preparation and after 3, 6 and 12 months of storage. FTIR and DSC studies were also carried out at the same time points. The ideal storage condition for the master dispersion was found to be at 5 degrees C. The addition of MCC, mannitol and lactose (alpha-monohydrate) to the original dispersion afforded a solidification effect on the formulation at room temperature and showed the same dissolution behavior (not less than 85% of piroxicam within 30 min in pH 1.2, 4.5 and 6.8; and water) with the master. The dispersion including lactose was stable at 25 degrees C for 12 months. However, the ideal period of storage for the modified dispersions including MCC and mannitol was 6 months at 25 degrees C. FTIR and DSC results both confirmed the amorphous state of piroxicam in all semi-solid dispersions under storage conditions for 12 months.
Product Description
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Mannitol, European Pharmacopoeia (EP) Reference Standard
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Gelatin from bovine skin, gel strength ~225 g Bloom, Type B
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Sigma-Aldrich
Gelatin, tested according to Ph. Eur.
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Gelatin from porcine skin, Type A, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Gelatin from bovine skin, Type B
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Piroxicam for system suitability, European Pharmacopoeia (EP) Reference Standard
Piroxicam, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Gelatin from bovine skin, Type B, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Prionex® Highly purified Type A, aqueous solution
Sigma-Aldrich
Piroxicam, meets USP testing specifications
Sigma-Aldrich
Gelatin from cold water fish skin, solid
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A
Sigma-Aldrich
Gelatin from porcine skin, gel strength ~175 g Bloom, Type A
Millipore
Gelatin from porcine skin, suitable for microbiology, high gel strength
Millipore
Gelatin from porcine skin, suitable for microbiology, low gel strength
Millipore
Gelatin from porcine skin, medium gel strength, suitable for microbiology
Sigma-Aldrich
Lactose, tested according to Ph. Eur.
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture