Merck
  • Combination of targeted PDT and anti-VEGF therapy for rat CNV by RGD-modified liposomal photocyanine and sorafenib.
Combination of targeted PDT and anti-VEGF therapy for rat CNV by RGD-modified liposomal photocyanine and sorafenib.
Investigative ophthalmology & visual science (2013-11-10)
Jia-lin Wang, Yi Xi, Yu-ling Liu, Zhao-hui Wang, Qiang Zhang
ABSTRACT
To achieve a combination therapy of targeted PDT and anti-VEGF for choroidal neovascularization (CNV). Arg-Gly-Asp (RGD)-modified liposomes encapsulating photocyanine and sorafenib (RGD-SSL-[P]-[S]) were prepared and characterized. Drug concentration in RGD-SSL-[P]-[S] and irradiation time were optimized on ARPE-19 and HUVEC cells in vitro. A laser-induced CNV rat model was used to assess the efficacy of this targeted combinational system. The effect of RGD-SSL-[P]-[S] on the retinal structure of BN rats was also examined. The particle size of RGD-SSL-[P]-[S] was approximately 100 nm, with a encapsulation efficiency more than 90% for both photocyanine and sorafenib. From RGD-SSL-[P]-[S], the release rate of photocyanine was approximately 22%, whereas that of sorafeinb was approximately 40% at 48 hours. With the optimal drug concentration and irradiation time, RGD-SSL-[P]-[S] exhibited cytotoxicity only to HUVEC without obvious damage to normal ARPE-19 cells. Rats treated with RGD-SSL-[P]-[S] showed the least CNV area and fluorescein leakage in fluorescein fundus angiography. RGD-SSL-[P]-[S] was also found safe to the rat retina. Combination of targeted PDT and anti-VEGF might be an effective therapy for CNV.
Product Description
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Niacinamide, meets USP testing specifications