Merck
Activation of a unique flavin-dependent tRNA-methylating agent.
Biochemistry (2013-11-16)
Djemel Hamdane, Eduardo Bruch, Sun Un, Martin Field, Marc Fontecave
ABSTRACT
TrmFO is a tRNA methyltransferase that uses methylenetetrahydrofolate (CH2THF) and flavin adenine dinucleotide hydroquinone as cofactors. We have recently shown that TrmFO from Bacillus subtilis stabilizes a TrmFO-CH2-FADH adduct and an ill-defined neutral flavin radical. The adduct contains a unique N-CH2-S moiety, with a methylene group bridging N5 of the isoalloxazine ring and the sulfur of an active-site cysteine (Cys53). In the absence of tRNA substrate, this species is remarkably stable but becomes catalytically competent for tRNA methylation following tRNA addition using the methylene group as the source of methyl. Here, we demonstrate that this dormant methylating agent can be activated at low pH, and we propose that this process is triggered upon tRNA addition. The reaction proceeds via protonation of Cys53, cleavage of the C-S bond, and generation of a highly reactive [FADH(N5)═CH2]+ iminium intermediate, which is proposed to be the actual tRNA-methylating agent. This mechanism is fully supported by DFT calculations. The radical present in TrmFO is characterized here by optical and EPR/ENDOR spectroscopy approaches together with DFT calculations and is shown to be the one-electron oxidized product of the TrmFO-CH2-FADH adduct. It is also relatively stable, and its decomposition is facilitated by high pH. These results provide new insights into the structure and reactivity of the unique flavin-dependent methylating agent used by this class of enzymes.
Sigma-Aldrich
Ribonucleic acid, transfer from Escherichia coli, Type XX, Strain W, lyophilized powder
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), buffered aqueous solution
Sigma-Aldrich
Ribonucleic acid, transfer from bovine liver, Type XI, lyophilized powder
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), buffered aqueous solution
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), Type X-SA, lyophilized powder
Sigma-Aldrich
Ribonucleic acid, transfer from wheat germ, Type V, 15-19 units/mg solid