Merck
  • Novel role of ZmaNAC36 in co-expression of starch synthetic genes in maize endosperm.
Novel role of ZmaNAC36 in co-expression of starch synthetic genes in maize endosperm.
Plant molecular biology (2013-11-16)
Junjie Zhang, Jiang Chen, Qiang Yi, Yufeng Hu, Hanmei Liu, Yinghong Liu, Yubi Huang
ABSTRACT
Starch is an essential commodity that is widely used as food, feed, fuel and in industry. However, its mechanism of synthesis is not fully understood, especially in terms of the expression and regulation of the starch synthetic genes. It was reported that the starch synthetic genes were co-expressed during maize endosperm development; however, the mechanism of the co-expression was not reported. In this paper, the ZmaNAC36 gene was amplified by homology-based cloning, and its expression vector was constructed for transient expression. The nuclear localization, transcriptional activation and target sites of the ZmaNAC36 protein were identified. The expression profile of ZmaNAC36 showed that it was strongly expressed in the maize endosperm and was co-expressed with most of the starch synthetic genes. Moreover, the expressions of many starch synthesis genes in the endosperm were upregulated when ZmaNAC36 was transiently overexpressed. All our results indicated that NAC36 might be a transcription factor and play a potential role in the co-expression of starch synthetic genes in the maize endosperm.
Sigma-Aldrich
Starch from potato, Powder
Sigma-Aldrich
Starch from wheat, Unmodified
Sigma-Aldrich
Starch from potato, suitable for electrophoresis
Sigma-Aldrich
Starch from potato, Soluble
Sigma-Aldrich
Starch, puriss. p.a., from potato, reag. ISO, reag. Ph. Eur., soluble
Sigma-Aldrich
Starch, from potato, tested according to Ph. Eur.
Supelco
Starch from corn, for use with Total Dietary Fiber Control Kit, TDF-C10
Supelco
Starch from corn, analytical standard, analytical standard for Starch Assay Kits SA-20 and STA-20
Sigma-Aldrich
Starch from corn, practical grade
Sigma-Aldrich
Starch from corn
Sigma-Aldrich
Starch from rice
Sigma-Aldrich
Starch from corn