Merck
  • Preparation and characterization of graphene oxide encapsulated gold nanoparticles.
Preparation and characterization of graphene oxide encapsulated gold nanoparticles.
Journal of nanoscience and nanotechnology (2013-11-20)
Yong Ju Yun, Ki-Bong Song
ABSTRACT
We present a simple approach for the fabrication of graphene oxide-encapsulated gold nanoparticles using graphene oxide sheet-wrapping via electrostatic self-assembly. By mixing bovine serum albumin molecule-functionalized gold nanoparticles with graphene oxide dispersion, positively charged bovine serum albumin/gold nanoparticles easily assembled with negatively charged graphene oxide sheets through electrostatic interaction. Transmittance electron microscopy, scanning electron microscopy, atomic force microscopy, and Raman spectroscopy were used to confirm the encapsulation of graphene oxide on gold nanoparticles. Interestingly, graphene oxide sheets wrapping mainly occurs along the main body of single or a few gold nanoparticles. Additionally, by measuring the ultraviolet-visible spectroscopy spectrum, we found that the surface plasmon resonances band of the graphene oxide-encapsulated gold nanoparticles was found to become red-shifted compared to that of pristine gold nanoparticles, whereas similar to that of bovine serum albumin-coated gold nanoparticles. These results indicating that most of graphene oxide-encapsulated gold nanoparticles have good monodispersity and spherical shape. These resulting materials may potentially serve as a platform for plasmon resonance electron transfer spectroscopy or a probe for low level biosensing.
Sigma-Aldrich
Gold, evaporation slug, diam. × L 0.3 cm × 0.6 cm, 99.99% trace metals basis
Sigma-Aldrich
Gold, powder, <850 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.025 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.5 mm, 99.999% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold coated glass slide, layer thickness 100 Å, 99.999% (Au)
Sigma-Aldrich
Gold coated glass cover slip, 99.999% (Au), diam. × thickness 15 mm × 130-170 μm, layer thickness 100 Å
Sigma-Aldrich
Gold coated glass cover slip, L × W × thickness 22 mm × 22 mm × 130-170 μm, square, layer thickness 100 Å, 99.999% (Au)
Sigma-Aldrich
Gold coated glass slide, layer thickness 1000 Å, 99.999% (Au)
Sigma-Aldrich
Gold, nanopowder, <100 nm particle size, 99.9% trace metals basis
Sigma-Aldrich
Graphite, powder, <45 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Graphite, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Gold, powder, <45 μm, 99.99% trace metals basis
Sigma-Aldrich
Gold, rod, diam. 3.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Graphite, rod, L 150 mm, diam. 3 mm, low density, 99.995% trace metals basis
Sigma-Aldrich
Graphite, rod, L 150 mm, diam. 6 mm, 99.995% trace metals basis
Sigma-Aldrich
Gold, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.05 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Graphite, powder, <20 μm, synthetic
Sigma-Aldrich
Gold, beads, 1-6 mm, 99.999% trace metals basis
Sigma-Aldrich
Gold, powder, <10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Graphite, flakes
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, ≥99.9% trace metals basis