Merck
  • Control of the visible emission in the SrZrO3 nano-crystals with the rare earth ion doping.
Control of the visible emission in the SrZrO3 nano-crystals with the rare earth ion doping.
Journal of nanoscience and nanotechnology (2013-11-20)
Dong Hwan Kim, Ji Hyun Kim, Jin Seok Chung, Yunsang Lee
ABSTRACT
We investigated the emission property of SrZrO3 nano-crystals (NCs) with the doping of rare earth (RE) ions, Eu3+ and Tm3+, by using 325 nm photo-excitation. SrZrO3 NCs show a sizable violet-blue emission, while the Eu3+ and Tm3+ ions are well known to be good red and blue phosphors, respectively. Combined emissions of the host and the RE ion dopant might suggest a new white luminescent source. The RE ion doped SrZrO3 NCs were initially synthesized by using the combustion method, and then the as-synthesized crystals were annealed at different temperatures from 650 degrees C to 1450 degrees C. The Eu3+-doped SrZrO3 NCs showed the sharp red emission near 600 nm, in addition to a violet-blue emission of the host material in itself. While the red emission is enhanced in the high temperature post-annealing, the blue emission is suppressed in an opposite way. This close relation between the emissions of the host and dopant was observed similarly in the Tm3+-doped NCs. We could control the emission property in the SrZrO3:Eu3+/Tm3+ NCs from blue to red by thermal annealing and RE ion doping.
Sigma-Aldrich
Strontium, granular, 99% trace metals basis
Sigma-Aldrich
Zirconium(IV) oxide, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Zirconium(IV) oxide, powder, 5 μm, 99% trace metals basis
Sigma-Aldrich
Strontium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Zirconium, powder, −100 mesh
Sigma-Aldrich
Zirconium(IV) oxide, 99.99% trace metals basis (purity excludes ~2% HfO2)
Sigma-Aldrich
Zirconium, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Strontium, random pieces, 99%
Sigma-Aldrich
Zirconium, sponge, ≥99% trace metals basis
Sigma-Aldrich
Strontium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Zirconium, rod, diam. 6.35 mm, ≥99% trace metals basis
Sigma-Aldrich
Zirconium(IV) oxide, nanoparticles, dispersion, <100 nm particle size (BET), 10 wt. % in H2O
Sigma-Aldrich
Zirconium(IV) oxide, nanoparticles, dispersion, <100 nm particle size (BET), 5 wt. % in H2O