Merck
Growth mechanism of cubic MgO granule via common ion effect.
Journal of nanoscience and nanotechnology (2013-11-20)
Se-Bum Choi, Nam-Woon Kim, Dong-Kyu Lee, Hyunung Yu
ABSTRACT
MgO is a typical wide bandgap insulator with its excellent thermodynamic stability, low dielectric constant, and low refractive index for growing various thin film materials. For this application, it is worth developing and understanding suitable growth method for MgO nanostructure. In this study, cubic MgCO3 have been successfully prepared via hydrolysis control of magnesium salt and alkaline solution with common ions to form nanoflake assembly. An optimum hydrothermal process and its shape evolution of cubic MgCO3 and MgO granules are analyzed. The physicochemical properties of the obtained samples are characterized by scanning electron microscopy (SEM), X-ray diffraction (XRD) and Fourier Transform InfraRed (FT-IR) to show high purity and structural uniformity of magnesium compounds.
Sigma-Aldrich
Magnesium oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 530, light, 98.0-100.5% (calc. for dried substance)
Sigma-Aldrich
Magnesium oxide, nanopowder, ≤50 nm particle size (BET)
Sigma-Aldrich
Magnesium oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, 99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, BioUltra, ≥97.0% (calcined substance, KT)
Sigma-Aldrich
Magnesium oxide, tested according to Ph. Eur., heavy
Sigma-Aldrich
Magnesium oxide, (single crystal substrate), ≥99.9% trace metals basis, <100>, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Magnesium oxide, light, 95%
Sigma-Aldrich
Magnesium oxide, -10-+50 mesh, 98%
Sigma-Aldrich
Magnesium oxide, ACS reagent, 97%
Sigma-Aldrich
Magnesium oxide, ≥99% trace metals basis, -325 mesh