Merck
  • [Cellulolytic activity of Fenellia flavipes and Fusarium oxysporum strains].
[Cellulolytic activity of Fenellia flavipes and Fusarium oxysporum strains].
Mikrobiolohichnyi zhurnal (Kiev, Ukraine : 1993) (2014-01-24)
T P Chepchak, S V Olishevskaia, I N Kurchenko
ABSTRACT
The production of cellulolytic enzymes by Fenellia flavipes and Fusarium oxysporum strains in submerged culture with plant residues as carbon source was studied. It was established that the majority of studied strains was able to hydrolyze the filter paper, husk of sunflower seeds, wheat straw and corn stalks. The ability to synthesize enzymes depended on the strain of microscopic fungi, type of substrate and duration of cultivation. As a result two fungal strains were selected: F. flavipes 655 with maximum of cellulolytic activity 2 U/ml in the medium with wheat straw and 1.6 U/ml in the medium with corn stalks on the 4th day of cultivation and F. oxysporum 420 which synthesized 0.875 mg/ml of reducing sugars.
Sigma-Aldrich
Cellulose, microcrystalline, powder
Sigma-Aldrich
Cellulose, colloidal, microcrystalline
Sigma-Aldrich
Cellulose, microcrystalline, powder, 20 μm
Supelco
Cellulose, acid washed, from spruce, suitable for column chromatography
Supelco
Cellulose, DS-0, powder, suitable for thin layer chromatography (TLC)
Supelco
Cellulose, powder, suitable for column chromatography
Supelco
Cellulose, acid washed, powder, suitable for column chromatography
Supelco
Cellulose, DFS-0, microcrystalline, suitable for thin layer chromatography (TLC)
Sigma-Aldrich
Avicel® PH-101, tested according to Ph. Eur.
Sigma-Aldrich
Cellulose, fibers, (medium)
Sigma-Aldrich
α-Cellulose, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm
Supelco
Avicel® PH-101, ~50 μm particle size
Sigma-Aldrich
α-Cellulose, powder
Sigma-Aldrich
Sigmacell Cellulose, Type 101, Highly purified, fibers