Merck
  • Involvement of trypsin-digested silk peptides in the induction of RAW264.7 macrophage activation.
Involvement of trypsin-digested silk peptides in the induction of RAW264.7 macrophage activation.
Natural product communications (2014-02-22)
Kyoung-Ho Pyo, Min-Ki Kim, Kwang-Soon Shin, Hyang Sook Chun, Eun-Hee Shin
ABSTRACT
The activation of macrophages by trypsin-digested silk peptides was investigated by considering CD1 lb and CD40 expression in the RAW264.7 cell, a murine macrophage. Silk protein hydrolysates were digested with trypsin, following by centrifugal purification using the Centriprep 30k concentrator. Trypsin-digested total silk peptides and its centrifugal fractions were tested for macrophage activation in vitro. The functional peptide of fractionated silk peptides was examined by LC/MS/MS analysis. Trypsin-digested and fractionated silk peptides of more than 30 kDa induced an increase in the activation markers CD1 lb and CD40 in RAW264.7 cells. These results are supported by morphological changes reflecting an increase in the number of dendrites in activated cells. The fractionated silk peptides examined by LC/MS/MS contained partial peptides of Bombyx mori fibroin. These results suggest that the activation of RAW264.7 macrophages may be induced not by sericin-derived peptides but by fibroin-derived ones.
Sigma-Aldrich
Trypsin from bovine pancreas, powder, ≥7,500 BAEE units/mg solid
Sigma-Aldrich
Trypsin from bovine pancreas, essentially salt-free, lyophilized powder, ≥9,000 BAEE units/mg protein, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypsin from human pancreas, salt-free, lyophilized powder, vial of ≥1,000 BAEE units
Sigma-Aldrich
Trypsin Acetylated from bovine pancreas, Type V-S, ≥8,500 BAEE units/mg protein (biuret)
Sigma-Aldrich
Trypsin from bovine pancreas, Type I, ~10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, Type XI, lyophilized powder, ≥6,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, ≥2,500 USP units/mg solid, meets USP testing specifications
Sigma-Aldrich
Trypsin from porcine pancreas, ~1500 U/mg
Sigma-Aldrich
TrypZean® bovine, recombinant, expressed in corn, lyophilized powder, ≥3,350 units/mg solid (USP)
Sigma-Aldrich
Trypsin from porcine pancreas, tablet, 1 mg tablet
Sigma-Aldrich
Trypsin from bovine pancreas, suitable for protein sequencing, lyophilized powder
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, BioReagent, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, Type II-S, 1,000-2,000 units/mg dry solid
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
Sigma-Aldrich
Trypsin from porcine pancreas, Type IX-S, lyophilized powder, 13,000-20,000 BAEE units/mg protein