Merck
  • BTNL8, a butyrophilin-like molecule that costimulates the primary immune response.
BTNL8, a butyrophilin-like molecule that costimulates the primary immune response.
Molecular immunology (2013-09-17)
Andrei I Chapoval, Glennda Smithson, Linda Brunick, Mehdi Mesri, Ferenc L Boldog, David Andrew, Nikolai V Khramtsov, Elena A Feshchenko, Gary C Starling, Peter S Mezes
ABSTRACT
The role of the B7 family molecules in the regulation of the immune response is well documented. A large body of experimental evidence indicates that costimulatory molecules such as B7-1, B7-2, B7-DC, B7-H1, B7-H2, B7-H3 and B7-H4 are critical for initiation, maintenance and down-regulation of the immune response. However the immunological function of butyrophilin (BTN)-like molecules, which are a part of the expanded B7 family, is not known. Here, we demonstrate that the extracellular portion of human BTNL8 can augment Ag-induced activation of T lymphocytes. BTNL8 has two alternatively spliced forms: B7-like and BTN-like. Both isoforms of BTNL8 were expressed concurrently in various human tissues. A putative BTNL8 receptor was detected only on resting T lymphocytes. Administration of BTNL8Ig fusion protein into mice promoted production of Ag-specific IgG during the primary, but not the secondary immune responses. BTNL8 may therefore play an essential role in priming of naïve T lymphocytes.