Merck
  • Action of amorphous diatomaceous earth against different stages of the stored product pests Tribolium confusum, Tenebrio molitor, Sitophilus granarius and Plodia interpunctella.
Action of amorphous diatomaceous earth against different stages of the stored product pests Tribolium confusum, Tenebrio molitor, Sitophilus granarius and Plodia interpunctella.
Journal of stored products research (2000-12-22)
Mewis, Ulrichs
ABSTRACT
Environmental and human health problems associated with the use of synthetic pesticides have prompted the demand for non-polluting, biologically specific insecticides. The current study tested the use and action of diatomaceous earth against several stored product pests. Fossil Shield(R) applied to wooden plates was lethal to adult Tenebrio molitor and Tribolium confusum, but larvae of the mealworm were unaffected. Beetles died within 14 days exposure in the absence of food to a dose of 2 and 4 g/m(2), but mortality was reduced in those fed grain bran. Fossil Shield(R) was lethal to first instar larvae of Plodia interpunctella, but not lethal to older larval stages. Two-week old larvae of T. confusum were more sensitive to diatomaceous earth than P. interpunctella at the same age. Contact with diatomaceous earth caused adult Sitophilus granarius, T. molitor and T. confusum to lose weight and reduced their water content, suggesting disruption of "the water barrier". Death of stored product insects treated with diatomaceous earth decreased with increased r.h., due to reduced transpiration through the cuticle. High r.h. delays, or above 60% can prevent, the drying action of diatomaceous earth.
Product Description
Supelco
Celite® 545 AW, reagent grade
Pricing and availability is not currently available.