Merck
  • Water soluble sodium sulfate nanorods as a versatile template for the designing of copper sulfide nanotubes.
Water soluble sodium sulfate nanorods as a versatile template for the designing of copper sulfide nanotubes.
Journal of nanoscience and nanotechnology (2014-04-18)
Gautam Das, Nitul Kakati, Seok Hee Lee, Niranjan Karak, Young Soo Yoon
ABSTRACT
The present study reports the use of water soluble sodium sulfate (Na2SO4) nanorods as a versatile template for generation of tubular copper sulfide (CuS) nanostructures. The Na2SO4 nanorods were synthesized from ammonium sulfate (NH4)2SO4 and sodium hydroxide (NaOH), under refluxing condition. The shape and morphology control of the Na2SO4 nanorods were studied with respect to nature of surfactant used and reactant mole ratio. While, PVP mole ratio was important to obtain homogeneous nanorods. Uniform and stable nanotubes of CuS were than obtained by the dissolution of the nanorods in water. The use of simple chemicals for synthesis of such nanotube templates opens the prospect for wide scale downstream applications.
Product Description
Supelco
Water, for TOC analysis
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Sigma-Aldrich
Sodium hydrogen sulfate
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Supelco
Water, suitable for HPCE, suitable for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Water, BioPerformance Certified