Merck
  • Influence of mounthrinse solutions on the color stability and microhardness of a composite resin.
Influence of mounthrinse solutions on the color stability and microhardness of a composite resin.
The international journal of esthetic dentistry (2014-04-26)
Cesar Penazzo Lepri, Marcus Vinicius de Melo Ribeiro, Alessandro Dibb, Regina Guenka Palma-Dibb
ABSTRACT
The aim of the present study was to evaluate three mouth rinse solutions' (Plax, Listerine and Periogard) influence on the microhardness and color stability of a composite resin (Esthet.X, Dentsply). For this purpose, forty specimens were prepared, being 10 specimens for each solution and 10 for artificial saliva (control group). Initially, specimens were polished and immersed in artificial saliva and following the initial color, an evaluation was made. For 30 days, specimens were immersed in the solutions three times a day for 1 min. After this cycling period, color change was analyzed and Knoop microhardness measures were carried out. A new color measurement was performed after repolishing the specimens. The microhardness and color stability results showed no statistically significant difference among the studied solutions. However, samples immersed in Periogard showed a significant alteration in the composite resin brightness test. It can be concluded that mouth rinse can influence the luminosity of composite resin.
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Chlorhexidine digluconate solution, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Chlorhexidine digluconate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Triclosan, 97.0-103.0% (active substance, GC)
Sigma-Aldrich
Chlorhexidine dihydrochloride, ≥98%
Chlorhexidine dihydrochloride, European Pharmacopoeia (EP) Reference Standard
Chlorhexidine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Chlorhexidine, ≥99.5%
Supelco
Chlorhexidine, Pharmaceutical Secondary Standard; Certified Reference Material
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Supelco
Triclosan, Pharmaceutical Secondary Standard; Certified Reference Material