Merck
  • Proposal of a new degradation mechanism of enalapril maleate and improvement of enalapril maleate stability in tablet formulation with different stabilizers.
Proposal of a new degradation mechanism of enalapril maleate and improvement of enalapril maleate stability in tablet formulation with different stabilizers.
Die Pharmazie (2014-05-06)
J Chen, L H Zhang, R J Xu, N J Bu, L Zhang
ABSTRACT
Enalapril maleate (EM) is unstable in poorly designed tablet formulations. To improve the stability of EM, the degradation mechanism should be elucidated. In this study, we found that several commonly used excipients promoted the degradants of EM, particularly a diketopiperazine derivative (DKP). We propose two degradation pathways in which both acid and alkali can promote the formation of DKP, although previous reports suggested that DKP is produced mainly in acidic media. Based on the degradation pathways, we believe that subtle control of the microenvironmental pH can inhibit the formation of DKP. This was confirmed by the observation that the degradation rate became slower when certain organic acids were added to the binary mixtures of EM and excipient. The data showed that the stability of EM in the ternary mixtures was much higher than that in binary mixtures. It was further proved that tablets containing these organic acids produced less DKP after the accelerated test. We also found that the formation of DKP in tablets varied with different ratios of tartaric acid, which was used as a model organic acid. This illustrated that an optimum ratio of tartaric acid is required. These results indicated that the stability of EM in tablet formulation is closely associated with microenvironmental pH and the addition of a suitable organic acid based on the reaction mechanism is an effective strategy for improving the stability of EM.
Product Description
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Enalapril maleate salt, powder, ≥98% (HPLC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Enalapril Maleate, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Enalapril maleate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ultrapure Acetonitrile
Enalapril maleate, European Pharmacopoeia (EP) Reference Standard
Enalapril for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%