Merck
Lovastatin and beyond: the history of the HMG-CoA reductase inhibitors.
Nature reviews. Drug discovery (2003-06-20)
Jonathan A Tobert
ABSTRACT
In the 1950s and 1960s, it became apparent that elevated concentrations of plasma cholesterol were a major risk factor for the development of coronary heart disease, which led to the search for drugs that could reduce plasma cholesterol. One possibility was to reduce cholesterol biosynthesis, and the rate-limiting enzyme in the cholesterol biosynthetic pathway, 3-hydroxy-3-methyl-glutaryl-CoA (HMG-CoA) reductase, was a natural target. Here, I describe the discovery and development of lovastatin--the first approved inhibitor of HMG-CoA reductase--and the clinical trials that have provided the evidence for the ability of drugs in this class to reduce the morbidity and mortality associated with cardiovascular disease.
Product Description
USP
Lovastatin, United States Pharmacopeia (USP) Reference Standard
Supelco
Lovastatin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Mevinolin from Aspergillus sp., ≥98% (HPLC)
Lovastatin, European Pharmacopoeia (EP) Reference Standard
Lovastatin for peak identification, European Pharmacopoeia (EP) Reference Standard