Merck
  • Sporotrichosis successfully treated with terbinafine and potassium iodide: case report and review of the literature.
Sporotrichosis successfully treated with terbinafine and potassium iodide: case report and review of the literature.
Mycopathologia (2004-10-19)
Basak Coskun, Yunus Saral, Nusret Akpolat, Arzu Ataseven, Demet Ciçek
ABSTRACT
Sporotrichosis is rare in Turkey. We report a 40-year-old woman who had subcutaneous sporotrichosis caused by sporothrix schenckii that was successfully treated with terbinafine (250 mg, twice a day) for a period of 6 months. She received a saturated solution of potassium iodide orally for two months. Terbinafine and potassium iodide are suggested to be the agents of choice for treatment of subcutaneous sporotrichosis.
Sigma-Aldrich
Potassium iodide, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥99.0%
Sigma-Aldrich
Potassium iodide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium iodide, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium iodide, puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium iodide, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Potassium iodide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium iodide, suitable for plant cell culture
Sigma-Aldrich
Potassium iodide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium iodide, ReagentPlus®, 99%
Sigma-Aldrich
Gold etchant, standard