Merck
  • Should acyclovir be prescribed for immunocompetent children presenting with chickenpox?
Should acyclovir be prescribed for immunocompetent children presenting with chickenpox?
Archives of disease in childhood (2005-05-24)
D Harris, J Redhead
Product Description
Supelco
Acyclovir, Pharmaceutical Secondary Standard; Certified Reference Material
Aciclovir, European Pharmacopoeia (EP) Reference Standard
USP
Acyclovir, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acycloguanosine, ≥99% (HPLC), powder