Merck
  • Dietary stearic acid and risk of cardiovascular disease: intake, sources, digestion, and absorption.
Dietary stearic acid and risk of cardiovascular disease: intake, sources, digestion, and absorption.
Lipids (2006-02-16)
Penny M Kris-Etherton, Amy E Griel, Tricia L Psota, Sarah K Gebauer, Jun Zhang, Terry D Etherton
ABSTRACT
Individual FA have diverse biological effects, some of which affect the risk of cardiovascular disease (CVD). In the context of food-based dietary guidance designed to reduce CVD risk, fat and FA recommendations focus on reducing saturated FA (SFA) and trans FA (TFA), and ensuring an adequate intake of unsaturated FA. Because stearic acid shares many physical properties with the other long-chain SFA but has different physiological effects, it is being evaluated as a substitute for TFA in food manufacturing. For stearic acid to become the primary replacement for TFA, it is essential that its physical properties and biological effects be well understood.
Product Description
Supelco
Stearic acid, analytical standard
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Supelco
Stearic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Supelco
Stearic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Stearic acid, United States Pharmacopeia (USP) Reference Standard