Merck
  • Use of high-dose epinephrine and sodium bicarbonate during neonatal resuscitation: is there proven benefit?
Use of high-dose epinephrine and sodium bicarbonate during neonatal resuscitation: is there proven benefit?
Clinics in perinatology (2006-03-15)
Myra H Wyckoff, Jeffrey M Perlman
ABSTRACT
For adults and pediatric age patients, high-dose intravenous epinephrine was recommended if standard-dose epinephrine failed to achieve return of spontaneous circulation. More recent trials suggest that high-dose epinephrine is not beneficial and may result in increased harm. There are no randomized clinical studies of high-dose versus standard-dose intravenous epinephrine in neonates. Routine use of high-dose epinephrine during neonatal resuscitation cannot be recommended. Although sodium bicarbonate has been used during neonatal resuscitation, the only randomized controlled trial of its use during brief neonatal resuscitation showed no benefit. Sodium bicarbonate infusion during neonatal cardiopulmonary resuscitation (CPR) has several known and potential side effects. The use of sodium bicarbonate infusion should be discouraged during brief CPR. Whether sodium bicarbonate is beneficial for infants who require prolonged CPR despite adequate ventilation is unknown.
Product Description
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate solution, solution (7.5%), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Supelco
Sodium bicarbonate concentrate, 0.1 M NaHCO3 in water, eluent concentrate for IC
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C