- [Comparative evaluation of the safety and efficacy of etoricoxib and diclofenac on the upper gastrointestinal tract in patients with osteoarthrosis and rheumatoid arthritis (the multinational etoricoxib and diclofenac arthritis long-term (MEDAL) study program)].
[Comparative evaluation of the safety and efficacy of etoricoxib and diclofenac on the upper gastrointestinal tract in patients with osteoarthrosis and rheumatoid arthritis (the multinational etoricoxib and diclofenac arthritis long-term (MEDAL) study program)].
Terapevticheskii arkhiv (2010-09-29)
L I Alekseeva
PMID20873248
ABSTRACT
The review gives and analyzes the data of a number of studies evaluating the safety and efficacy of nonsteroidal anti-inflammatory drugs on the upper gastrointestinal tract. Particular emphasis is laid on the results of the multinational etoricoxib and diclofenac arthritis long-term (MEDAL) study program. The randomized MEDAL study has shown that etoricoxib has more benefits than diclofenac.