Merck
  • Tungsten-rhenium suture needles with improved properties for coronary artery bypass graft surgery.
Tungsten-rhenium suture needles with improved properties for coronary artery bypass graft surgery.
Journal of biomedical materials research. Part B, Applied biomaterials (2010-06-25)
Frank R Cichocki, Robert E Maurer, S Neil Bar
ABSTRACT
Suture needles are essential instruments for performing blood vessel anastomosis in coronary artery bypass graft (CABG) surgeries. Exceptional needle performance is needed now as never before because of the increasing prevalence of elderly patients with advanced stages of coronary disease and calcified tissue. The various properties that affect the performance of suture needles used in CABG surgery are reviewed and recent progress towards improving needle performance through the use of tungsten-rhenium alloys and novel lubricous needle coatings is presented. Substantial enhancement of properties beyond those exhibited by commercially available stainless steel suture needles has been made, including an approximate 40% increase in strength, 100% increase in stiffness, and superior multiple pass penetration performance in both synthetic rubber media and human cadaver carotid arteries (p < 0.01).
Sigma-Aldrich
Tungsten, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Tungsten, wire, diam. 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tungsten, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tungsten, foil, thickness 0.5 mm, ≥99.9% trace metals basis
Tungsten, foil, not light tested, 100x100mm, thickness 0.015mm, 99.95%
Tungsten, wire, straight, 29mm, diameter 0.5mm, 99.9+%
Tungsten, wire, straight, 500mm, diameter 0.38mm, 99.9+%
Tungsten, wire reel, 5m, diameter 0.08mm, gold plated, 99.95%
Tungsten, wire, straight, 300mm, diameter 0.38mm, 99.9+%
Tungsten, wire reel, 5m, diameter 0.08mm, clean, 99.95%
Tungsten, foil, 0.2m coil, thickness 0.076mm, coil width 1.5mm, as rolled, 99.95%
Tungsten, foil, 0.5m coil, thickness 0.076mm, coil width 1.5mm, as rolled, 99.95%
Tungsten, foil, 1m coil, thickness 0.2mm, coil width .6mm, as rolled, 99.95%
Tungsten, foil, 0.2m coil, thickness 0.08mm, coil width .9mm, as rolled, 99.95%
Tungsten, foil, 50x50mm, thickness 3.0mm, as rolled, 99.95%
Tungsten, foil, 1m coil, thickness 0.025mm, coil width .7mm, as rolled, 99.95%
Tungsten, foil, 1m coil, thickness 0.05mm, coil width 2mm, as rolled, 99.95%
Tungsten, foil, 0.5m coil, thickness 0.076mm, coil width 2mm, as rolled, 99.95%
Tungsten, foil, 0.5m coil, thickness 0.02mm, coil width .5mm, as rolled, 99.95%
Tungsten, foil, 2m coil, thickness 0.1mm, coil width .6mm, as rolled, 99.95%
Tungsten, foil, 1m coil, thickness 0.05mm, coil width .51mm, as rolled, 99.95%
Tungsten, foil, 0.5m coil, thickness 0.025mm, coil width 1mm, as rolled, 99.95%
Tungsten, foil, 2m coil, thickness 0.035mm, coil width .5mm, as rolled, 99.95%
Tungsten, foil, 0.2m coil, thickness 0.02mm, coil width .5mm, as rolled, 99.95%
Tungsten, foil, 1m coil, thickness 0.05mm, coil width 1.5mm, as rolled, 99.95%
Tungsten, foil, 0.2m coil, thickness 0.035mm, coil width .75mm, as rolled, 99.95%
Tungsten, foil, 200x200mm, thickness 0.1mm, as rolled, 99.95%
Tungsten, foil, 1m coil, thickness 0.076mm, coil width 1.5mm, as rolled, 99.95%
Tungsten, foil, 50x50mm, thickness 2.0mm, as rolled, 99.95%
Tungsten, foil, 2m coil, thickness 0.025mm, coil width .7mm, as rolled, 99.95%