Merck
  • Fomepizole for the treatment of pediatric ethylene and diethylene glycol, butoxyethanol, and methanol poisonings.
Fomepizole for the treatment of pediatric ethylene and diethylene glycol, butoxyethanol, and methanol poisonings.
Clinical toxicology (Philadelphia, Pa.) (2010-07-01)
Jeffrey Brent
ABSTRACT
The use and clinical efficacy of the alcohol dehydrogenase inhibitor fomepizole is well established for the treatment of ethylene glycol and methanol poisonings in adults. A computerized search of the U.S. National Academy of medicine and EMBase databases was undertaken to identify published cases of patients treated with fomepizole. This search strategy identified 14 published cases related to the topic of this review: 10 due to ethylene glycol poisoning, 1 due to diethylene glycol poisoning, 1 due to butoxyethanol ingestion, and 2 due to methanol poisoning. The median age of these cases was 5.5 years old. FOMEPIZOLE IN GLYCOL AND GLYCOL ETHER POISONING: For the 10 ethylene glycol poisoned patients, the median recorded values of their arterial pH was 7.27 (range 7.03-7.38), serum bicarbonate concentration was 13 mEq/L (range 2-25), and ethylene glycol concentration was 2,140 mg/L (range 130-3,840). Eight of these patients were not hemodialyzed. The eight patients who were not hemodialyzed had ethylene glycol concentrations as high as 3,500 mg/L and serum bicarbonate concentrations as low as 4 mEq/L. All 10 patients had resolution of their metabolic acidosis and recovered without sequelae. The half-times of ethylene glycol elimination ranged from 9 to 15 h during fomepizole therapy, which is faster than the 19.7 h reported in adults. The two patients who ingested diethylene glycol or butoxyethanol all recovered without sequelae. The patient who ingested the butoxyethanol had a serum bicarbonate concentration of 13 mEq/L and was not hemodialyzed. FOMEPIZOLE IN METHANOL POISONING: One of the two children who ingested methanol was hemodialyzed. Both cases had a similar degree of severity. DOES FOMEPIZOLE OBVIATE THE NEED FOR HEMODIALYSIS?: Based on the experience reviewed herein it appears that, as in adults, hemodialysis may not be necessary in most cases of pediatric ethylene glycol poisoning if treated with fomepizole. FOMEPIZOLE PHARMACOKINETICS: Plasma fomepizole concentrations were measured in three cases and were found to be therapeutic with apparent Michaelis-Menton kinetics, having a zero-order elimination rate of 0.6-1 mg/L/h at higher concentrations and a first-order elimination with an apparent elimination half-time of 3.9 h at lower concentrations. FOMEPIZOLE REGIMEN: Most cases used the current U.S.-approved regimen. ADVERSE EFFECTS OF FOMEPIZOLE: The one adverse effect reported during fomepizole therapy was transient nystagmus in a 6-year-old with a serum ethylene glycol concentration of 130 mg/L and a serum bicarbonate concentration of 2 mEq/L; it is likely that ethylene glycol itself was the cause. COMPARISON OF FOMEPIZOLE WITH ETHANOL THERAPY: Two cases were originally treated with ethanol but switched to fomepizole because of adverse effects. In both cases, the adverse reactions to ethanol resolved once fomepizole treatment was initiated. The limited data available suggest that fomepizole, using the same dosage regimen as that used for adults, is efficacious and well tolerated in pediatric patients. In many cases of pediatric ethylene glycol poisoning treated with fomepizole, hemodialysis may not be necessary despite high concentrations and the presence of metabolic acidosis.
Sigma-Aldrich
Diethylene glycol, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
Diethylene glycol, puriss. p.a., ≥99.0% (GC), colorless
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Diethylene glycol, ReagentPlus®, 99%
Supelco
Methanol, analytical standard
Supelco
Diethylene glycol, analytical standard
USP
Diethylene glycol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Diethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Fomepizole, 99%
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard