Merck
  • Delayed neurological sequelae from ethylene glycol, diethylene glycol and methanol poisonings.
Delayed neurological sequelae from ethylene glycol, diethylene glycol and methanol poisonings.
Clinical toxicology (Philadelphia, Pa.) (2011-01-05)
Nandi J Reddy, Madhuri Sudini, Lionel D Lewis
ABSTRACT
Ethylene glycol, diethylene glycol and methanol are widely available chemicals and are found in a variety of common household products including antifreeze, windshield washer fluid, brake fluid and lubricants. Following ingestion of these glycols and methanol, patients frequently develop an early neurological syndrome consisting of inebriation, ataxia, and if severe, seizures and coma. Though uncommon, a neurological syndrome may also develop as a delayed complication. Using Pub Med 438 references were identified of which 45 were relevant. Ethylene glycol poisoning has produced cranial nerve deficits (usually VII nerve dysfunction) after a delay of 5-20 days, Parkinsonism and cerebral edema. Diethylene glycol ingestion has been associated with the development of optic nerve injury, cranial nerve deficits, quadraparesis and peripheral neuropathy. Methanol poisoning has led to Parkinsonism and polyneuropathy. Oxalate crystal deposition likely causes the cranial neuropathies related to ethylene glycol and 2-hydroxyethoxyacetic acid is thought to be the causal moiety in cranial neuropathies resulting from diethylene glycol toxicity. Formic acid is implicated in the optic nerve damage associated with methanol. Uncommonly, delayed neurological syndromes may develop as complications of poisoning due to ethylene glycol, diethylene glycol and methanol; the onset of such neurological damage is often days or even weeks post-ingestion. Further research is required to explain why the facial nerve is the cranial nerve most commonly involved and why the basal ganglia are predisposed to injury.
Product Description
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Diethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Ethylene glycol solution, suitable for NMR (reference standard), 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Supelco
Diethylene glycol, analytical standard
Sigma-Aldrich
Diethylene glycol, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
Diethylene glycol, puriss. p.a., ≥99.0% (GC), colorless
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Diethylene glycol, ReagentPlus®, 99%
USP
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethylene glycol, analytical standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
USP
Diethylene glycol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%