Merck
  • Metabolic engineering strategies for improving xylitol production from hemicellulosic sugars.
Metabolic engineering strategies for improving xylitol production from hemicellulosic sugars.
Biotechnology letters (2013-07-25)
Buli Su, Mianbin Wu, Jianping Lin, Lirong Yang
ABSTRACT
Xylitol is a five-carbon sugar alcohol with potential for use as a sweetener. Industrially, xylitol is currently produced by chemical hydrogenation of D-xylose using Raney nickel catalysts and this requires expensive separation and purification steps as well as high pressure and temperature that lead to environmental pollution. Highly efficient biotechnological production of xylitol using microorganisms is gaining more attention and has been proposed as an alternative process. Although the biotechnological method has not yet surpassed the advantages of chemical reduction in terms of yield and cost, various strategies offer promise for the biotechnological production of xylitol. In this review, the focus is on the most recent developments of the main metabolic engineering strategies for improving the production of xylitol.
Sigma-Aldrich
Xylitol, ≥99% (GC)
Supelco
Xylitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Xylitol
Supelco
Xylitol, analytical standard
Xylitol, European Pharmacopoeia (EP) Reference Standard