Merck
  • Effects of pharmacologic dopamine β-hydroxylase inhibition on cocaine-induced reinstatement and dopamine neurochemistry in squirrel monkeys.
Effects of pharmacologic dopamine β-hydroxylase inhibition on cocaine-induced reinstatement and dopamine neurochemistry in squirrel monkeys.
The Journal of pharmacology and experimental therapeutics (2014-05-13)
Debra A Cooper, Heather L Kimmel, Daniel F Manvich, Karl T Schmidt, David Weinshenker, Leonard L Howell
ABSTRACT
Disulfiram has shown promise as a pharmacotherapy for cocaine dependence in clinical settings, although it has many targets, and the behavioral and molecular mechanisms underlying its efficacy are unclear. One of many biochemical actions of disulfiram is inhibition of dopamine β-hydroxylase (DBH), the enzyme that converts dopamine (DA) to norepinephrine (NE) in noradrenergic neurons. Thus, disulfiram simultaneously reduces NE and elevates DA tissue levels in the brain. In rats, both disulfiram and the selective DBH inhibitor nepicastat block cocaine-primed reinstatement, a paradigm which is thought to model some aspects of drug relapse. This is consistent with some clinical results and supports the use of DBH inhibitors for the treatment of cocaine dependence. The present study was conducted to confirm and extend these results in nonhuman primates. Squirrel monkeys trained to self-administer cocaine were pretreated with disulfiram or nepicastat prior to cocaine-induced reinstatement sessions. Neither DBH inhibitor altered cocaine-induced reinstatement. Unexpectedly, nepicastat administered alone induced a modest reinstatement effect in squirrel monkeys, but not in rats. To investigate the neurochemical mechanisms underlying the behavioral results, the effects of DBH inhibition on extracellular DA were analyzed in the nucleus accumbens (NAc) using in vivo microdialysis in squirrel monkeys. Both DBH inhibitors attenuated cocaine-induced DA overflow in the NAc. Hence, the attenuation of cocaine-induced changes in accumbal DA neurochemistry was not associated with altered cocaine-seeking behavior. Overall, the reported behavioral effects of DBH inhibition in rodent models of relapse did not extend to nonhuman primates under the conditions used in the current studies.
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Tetraethylthiuram disulfide, ≥97.0% (S)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis