Merck
  • Acetylation regulates the stability of glutamate carboxypeptidase II protein in human astrocytes.
Acetylation regulates the stability of glutamate carboxypeptidase II protein in human astrocytes.
Biochemical and biophysical research communications (2014-06-19)
Ji-Young Choi, Jong-Ho Kim, Sangmee Ahn Jo
ABSTRACT
Glutamate carboxypeptidase II (GCPII) is known to be implicated in brain diseases such as schizophrenia and bipolar disorder, and dramatically increases in prostate cancer. Here, we investigated the regulation of GCPII expression in astrocytes and examined whether GCPII is epigenetically regulated through histone modification. In this study, valproic acid (VPA), a drug used for bipolar disorder and epilepsy and a known histone deacetylase (HDAC) inhibitor was used. We found that acute exposure of VPA for 4-6h increased the GCPII protein level in human astrocyte U87MG cells but did not have a similar effect after 12-24h exposure. Real-time polymerase chain reaction analysis revealed that VPA did not affect the GCPII mRNA expression. In contrast, decrease in GCPII protein level by cycloheximide treatment was blocked by VPA, indicating that VPA increases GCPII protein stability. Treatment with MG132, a proteasome inhibitor, suggested that the VPA-induced increase of GCPII protein level is dependent on the ubiquitin/proteasome pathway. In addition, immunoprecipitation analysis revealed that VPA increased the acetylation of GCPII protein at the lysine residues and facilitated a decrease of the poly-ubiquitinated GCPII level. Similarly, M344, a specific HDAC 1/6 inhibitor, also increased the GCPII protein level. In contrast, treatment with C646, a histone acetyltransferase inhibitor of p300/CBP, significantly reduced the level of GCPII protein. Taken together, this study demonstrated that the increase in GCPII induced by VPA is not due to the classical epigenetic mechanism, but via enhanced acetylation of lysine residues in GCPII.
Product Description
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Millipore
Cycloheximide solution, 0.1%, suitable for microbiology
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
M344, ≥98% (HPLC), powder
Supelco
Valproic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Sodium dodecyl sulfate, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Cycloheximide, PESTANAL®, analytical standard
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
USP
Valproic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
DL-Dithiothreitol solution, 1 M in H2O