Merck
  • Attributing population-scale human exposure to various source categories: merging exposure models and biomonitoring data.
Attributing population-scale human exposure to various source categories: merging exposure models and biomonitoring data.
Environment international (2014-06-18)
Hyeong-Moo Shin, Thomas E McKone, Deborah H Bennett
ABSTRACT
Information about the distribution of chemical-production mass with respect to use and release is a major and unavailable input for calculating population-scale exposure estimates. Based on exposure models and biomonitoring data, this study evaluates the distribution of total production volumes (and environmental releases if applicable) for a suite of organic compounds. We used Bayesian approaches that take the total intake from our exposure models as the prior intake distribution and the intake inferred from measured biomarker concentrations in the NHANES survey as the basis for updating. By carrying out a generalized sensitivity analysis, we separated the input parameters for which the modeled range of the total intake is within a factor of 2 of the intake inferred from biomonitoring data and those that result in a range greater than a factor of 2 of the intake. This analysis allows us to find the most sensitive (or important) parameters and the likelihood of emission rates for various source emission categories. Pie charts of contribution from each exposure pathway indicate that chemical properties are a primary determinant of the relative contribution of each exposure pathway within a given class of compounds. For compounds with relatively high octanol-water partition coefficients (Kow) such as di-2-ethylhexyl phthalate (DEHP), pyrene, 2,2',4,4'-tetrabromodiphenyl ether (PBDE-47), and 2,2',4,4',5,5'-hexabromodiphenyl ether (PBDE-153), more than 80% of exposure derives from outdoor food ingestion and/or indoor dust ingestion. In contrast, for diethyl phthalate (DEP), di-iso-butyl phthalate (DiBP), di-n-butyl phthalate (DnBP), butylbenzyl phthalate (BBP), and naphthalene, all relatively volatile compounds, either inhalation (indoor and outdoor) or dermal uptake from direct consumer use is the dominant exposure pathway. The approach of this study provides insights on confronting data gaps to improve population-scale exposure estimates used for high-throughput chemical prioritization.
Product Description
Supelco
Bis(2-ethylhexyl) phthalate solution, certified reference material, 2000 μg/mL in methanol
Plastic additive 01, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Diisobutyl phthalate, 99%
Sigma-Aldrich
Benzyl butyl phthalate, 98%
Sigma-Aldrich
Dioctyl phthalate, ≥99.5%
Supelco
Diisobutyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Bis(2-ethylhexyl) phthalate, Selectophore
Sigma-Aldrich
Dibutyl phthalate, 99%
Supelco
Dibutyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Benzyl butyl phthalate, analytical standard
Supelco
Dibutyl phthalate, PESTANAL®, analytical standard
Dibutyl phthalate, European Pharmacopoeia (EP) Reference Standard
Supelco
Bis(2-ethylhexyl) phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Benzyl butyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Bis(2-ethylhexyl) phthalate, PESTANAL®, analytical standard