Merck
  • Synthesis and photophysical studies of self-assembled multicomponent supramolecular coordination prisms bearing porphyrin faces.
Synthesis and photophysical studies of self-assembled multicomponent supramolecular coordination prisms bearing porphyrin faces.
Proceedings of the National Academy of Sciences of the United States of America (2014-07-01)
Yanhui Shi, Irene Sánchez-Molina, Changsheng Cao, Timothy R Cook, Peter J Stang
ABSTRACT
Multicomponent self-assembly, wherein two unique donor precursors are combined with a single metal acceptor instead of the more common two-component assembly, can be achieved by selecting Lewis-basic sites and metal nodes that select for heteroligated coordination spheres. Platinum(II) ions show a thermodynamic preference for mixed pyridyl/carboxylate coordination environments and are thus suitable for such designs. The use of three or more unique building blocks increases the structural complexity of supramolecules. Herein, we describe the synthesis and characterization of rectangular prismatic supramolecular coordination complexes (SCCs) with two faces occupied by porphyrin molecules, motivated by the search for new multichromophore complexes with promising light-harvesting properties. These prisms are obtained from the self-assembly of a 90° Pt(II) acceptor with a meso-substituted tetrapyridylporphyrin (TPyP) and dicarboxylate ligands. The generality of this self-assembly reaction is demonstrated using five dicarboxylate ligands, two based on a rigid central phenyl ring and three alkyl-spaced variants, to form a total of five free-base and five Zn-metallated porphyrin prisms. All 10 SCCs are characterized by (31)P and (1)H multinuclear NMR spectroscopy and electrospray ionization mass spectrometry, confirming the structure of each self-assembly and the stoichiometry of formation. The photophysical properties of the resulting SCCs were investigated revealing that the absorption and emission properties of the free-base and metallated porphyrin prisms preserve the spectral features associated with free TPyP.
Product Description
Zinc, foil, 10mm disks, thickness 1.0mm, as rolled, 99.95+%
Zinc, foil, 0.5m coil, thickness 0.075mm, as rolled, 99.95+%
Zinc, foil, 25mm disks, thickness 0.05mm, as rolled, 99.95+%
Zinc, foil, 10mm disks, thickness 0.02mm, 99.9%
Zinc, foil, 1m coil, thickness 0.125mm, as rolled, 99.95+%
Zinc, foil, 0.3m coil, thickness 0.175mm, as rolled, 99.95+%
Zinc, foil, 10mm disks, thickness 0.35mm, as rolled, 99.95+%
Zinc, foil, 0.5m coil, thickness 2.0mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.25mm, as rolled, 99.99+%
Zinc, foil, 0.5m coil, thickness 0.125mm, as rolled, 99.95+%
Zinc, foil, 10mm disks, thickness 0.25mm, as rolled, 99.99+%
Zinc, foil, 1m coil, thickness 0.175mm, as rolled, 99.95+%
Zinc, foil, 25mm disks, thickness 0.05mm, as rolled, 99.99+%
Zinc, foil, 25mm disks, thickness 0.125mm, as rolled, 99.99+%
Zinc, foil, 25mm disks, thickness 0.175mm, as rolled, 99.95+%
Zinc, foil, 1m coil, thickness 0.1mm, as rolled, 99.95+%
Zinc, foil, 10mm disks, thickness 0.125mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.125mm, as rolled, 99.99+%
Zinc, foil, 15mm disks, thickness 0.25mm, as rolled, 99.95+%
Zinc, foil, 0.5m coil, thickness 0.025mm, as rolled, 99.99+%
Zinc, foil, 1m coil, thickness 0.075mm, as rolled, 99.95+%
Zinc, foil, 0.5m coil, thickness 0.05mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.1mm, as rolled, 99.95+%
Zinc, foil, 10mm disks, thickness 0.0025mm, 99.9%
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn2+ in THF, highly reactive Rieke®metal
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis