Merck
  • Use of nanoporous Cu(II) ion imprinted polymer as a new sorbent for preconcentration of Cu(II) in water, biological, and agricultural samples and its determination by electrothermal atomic absorption spectrometry.
Use of nanoporous Cu(II) ion imprinted polymer as a new sorbent for preconcentration of Cu(II) in water, biological, and agricultural samples and its determination by electrothermal atomic absorption spectrometry.
Journal of AOAC International (2014-08-26)
Hamid Fazelirad, Mohammad Ali Taher, Hamid Ashkenani
ABSTRACT
A new and selective Cu(ll) ion imprinted polymer was prepared by formation of 1-(2-pyridylazo)-2-naphthol complex for selective extraction and preconcentration of Cu ions. Polymerization was performed with ethylene glycol dimethacrylate as a crosslinking monomer and methacrylic acid as a functional monomer in the presence of 2,2'-azobis(isobutyronitrile) as an initiator via a bulk polymerization method. The Cu(ll) imprinted polymeric particles were characterized by scanning electron microscopy and IR spectrometry. The imprint Cu ion was removed from the polymeric matrix using 2 M HNO3. Optimum pH for maximum sorption was 5-8. Maximum sorbent capacity and the enrichment factor for Cu(ll) were 11.3 mg/g and 100, respectively. The RSD and LOD of the method were evaluated as +/-4.3% and 8.7 ng/L, respectively. The proposed method is simple, highly selective, and sensitive and can be applied to the determination of ultratrace amounts of Cu in water, biological, and agricultural samples.
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Copper, wire reel, 200m, diameter 0.5mm, annealed, silver plated, 99.9%
Copper - O.F.H.C., foil, 10m coil, thickness 0.05mm, annealed, 99.95+%
Copper - O.F.H.C., foil, 50x50mm, thickness 0.1mm, hard, 99.95+%
Copper, insulated wire, 100m, conductor diameter 0.05mm, insulation thickness 0.002mm, 2 micron polyesterimide insulation, 99.9%
Copper - O.F.H.C., wire reel, 100m, diameter 0.75mm, as drawn, 99.95+%
Copper - O.F.H.C., foil, 100x100mm, thickness 0.1mm, half hard, 99.95+%
Copper - O.F.H.C., foil, 10m coil, thickness 0.025mm, hard, 99.95+%
Copper - O.F.H.C., foil, 10m coil, thickness 0.05mm, coil width 10mm, annealed, 99.95+%
Copper - O.F.H.C., foil, 0.5m coil, thickness 0.05mm, hard, 99.95+%
Copper - O.F.H.C., foil, 100x100mm, thickness 0.5mm, half hard, 99.95+%
Copper, insulated wire, 250m, conductor diameter 0.05mm, insulation thickness 0.002mm, 2 micron polyesterimide insulation, 99.9%
Copper - O.F.H.C., foil, 300x300mm, thickness 0.1mm, half hard, 99.95+%
Copper, wire reel, 100m, diameter 0.05mm, annealed, 99.9%
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Copper, wire reel, 5m, diameter 1.0mm, as drawn, 99.98+%
Copper - O.F.H.C., foil, 8mm disks, thickness 0.5mm, half hard, 99.95+%
Copper, wire reel, 5m, diameter 0.01mm, hard, 99.99%