Merck
  • Development of a stir bar sorptive extraction method for analysis of ochratoxin A in beer.
Development of a stir bar sorptive extraction method for analysis of ochratoxin A in beer.
Journal of AOAC International (2014-08-26)
Kieu T N Nguyen, Dojin Ryu
ABSTRACT
A new, simple, and sensitive method was developed for extraction of ochratoxin A (OTA) in beer combined with HPLC-fluorescence detector. Samples were extracted by stir bar sorptive extraction (SBSE) followed by liquid desorption using commercially available Twister EG-Silicone. The main parameters influencing SBSE, including phase ratio, extraction time, stirring speed, ionic strength, organic modifier, pH, temperature, desorption mode, desorption solvent, and desorption time were optimized. Under the optimal conditions, assay was performed on 4 mL of samples acidified at pH 1.5. The samples were extracted for 180 min at a stirring speed of 800 rpm followed by desorption of analyte using 1 mL methanol under sonication for 45 min. The extract was evaporated at 50 degrees C under a gentle nitrogen stream, then redissolved in 200 microL of methanol-water (50 + 50, v/v). After each use, the stir bar was cleaned by sonication in methanol for 30 min three times. The method provided good linearity of the calibration curve with coefficients of determination greater than 0.999. Recoveries of OTA were greater than 83% with RSD lower than 10%, and LOD of OTA in beer was 0.64 ng/L. This method was applied to determine OTA in 19 beer samples. OTA was detected in 12 samples (63%) in the range of 0.01-0.27 ng/mL.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Supelco
Methanol, analytical standard
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium chloride solution, 5 M