Merck
  • Comparison of TiO2 nanoparticle and graphene-TiO2 nanoparticle composite phototoxicity to Daphnia magna and Oryzias latipes.
Comparison of TiO2 nanoparticle and graphene-TiO2 nanoparticle composite phototoxicity to Daphnia magna and Oryzias latipes.
Chemosphere (2014-07-23)
Shibin Li, Xuan Pan, Lindsay K Wallis, Zhaoyang Fan, ZuLiang Chen, Stephen A Diamond
ABSTRACT
With a dramatic rise in complexity, needs of nanotoxicology research go beyond simple forms of nanomaterials. This study compared the phototoxicity of nano-TiO2 and graphene-TiO2 nanocomposite (GNP). GNP was synthesized based on a hydrothermal method, which simultaneously performed the reduction of graphene oxide and nano-TiO2 loading. A series of acute toxicity tests of nano-TiO2, graphene and GNP was performed on two aquatic organisms, Daphnia magna and Oryzias latipes. Fast and substantial agglomeration and sedimentation of nanoparticles in test media and surface attachment of nano-TiO2 and GNP on D. magna surface was observed. Similar phototoxicity of nano-TiO2 and GNP for both species existed, though compared with nano-TiO2, GNP had a 2.3-fold increase in visible light photocatalytic ROS generation. In summary, this study demonstrated the significance of illumination spectrum, particle behavior, and species sensitivity on nanophototoxicity, and the needs for research on increasingly sophisticated functional materials.
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Titanium, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, 99.8% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Titanium, ≤10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Sigma-Aldrich
Titanium(IV) oxide, rutile, 99.995% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.25 mm, 99.7% trace metals basis
Titanium, IRMM®, certified reference material, 0.5 mm foil
Titanium, rod, 100mm, diameter 9.5mm, annealed, 99.99+%
Titanium, rod, 100mm, diameter 2mm, as drawn, 99.99+%
Titanium, rod, 100mm, diameter 10mm, annealed, 99.6+%
Titanium, rod, 200mm, diameter 16mm, annealed, 99.6+%
Titanium, rod, 100mm, diameter 16mm, as drawn, 99.99+%
Titanium, wire reel, 5m, diameter 0.75mm, as drawn, 99.6+%
Titanium, rod, 200mm, diameter 9.5mm, annealed, 99.99+%
Titanium, rod, 25mm, diameter 50mm, 99.99+%
Titanium, tube, 1220mm, outside diameter 2.03mm, inside diameter 1.55mm, wall thickness 0.24mm, annealed, 99.6+%
Titanium, rod, 500mm, diameter 4mm, annealed, 99.6+%
Titanium, tube, 1000mm, outside diameter 6.35mm, inside diameter 4.57mm, wall thickness 0.89mm, annealed, 99.6+%
Titanium, tube, 200mm, outside diameter 25.4mm, inside diameter 23.62mm, wall thickness 0.89mm, annealed, 99.6+%
Titanium, rod, 500mm, diameter 6mm, annealed, 99.6+%
Titanium, tube, 1000mm, outside diameter 9.5mm, inside diameter 8.2mm, wall thickness 0.65mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 25.4mm, inside diameter 23.62mm, wall thickness 0.89mm, annealed, 99.6+%