Merck
  • Acetylated FoxO1 mediates high-glucose induced autophagy in H9c2 cardiomyoblasts: regulation by a polyphenol -(-)-epigallocatechin-3-gallate.
Acetylated FoxO1 mediates high-glucose induced autophagy in H9c2 cardiomyoblasts: regulation by a polyphenol -(-)-epigallocatechin-3-gallate.
Metabolism: clinical and experimental (2014-07-27)
Jia Liu, Ying Tang, Zhihui Feng, Chen Hou, Hui Wang, Jiong Yan, Jing Liu, Weili Shen, Weijin Zang, Jiankang Liu, Jiangang Long
ABSTRACT
FoxO1 acts as a pivotal transcription factor in insulin signaling. However, in hyperglycemia induced cardiac complications, whether FoxO1 is involved remains unclear. The goal of this study was to delineate the potential role of FoxO1 under high-glucose condition. We investigated insulin resistance and reactive oxygen species (ROS) generation in H9c2 cardiomyoblasts after high-glucose exposure. A series of autophagy biomarkers were measured and further confirmed by LC3 turnover assay. Using gene silencing and overexpression experiments we dissected the molecular mechanisms of FoxO1 regulated autophagy. We also tested the protective effect of (-)-epigallocatechin-3-gallate (EGCG, a green tea-derived polyphenol) in high-glucose treated H9c2 cardiomyoblasts. High-glucose elicited elevated ROS, autophagy and FoxO1 abundance in cultured H9c2 cardiomyoblasts. Specifically, high-glucose significantly augmented the acetylated FoxO1 in cytosol. In line, compared with 3A-FoxO1 (majorly localized in nuclei with a strong transcriptional activity), overexpression of WT-FoxO1 led to more intense elevated autophagy with enhanced acetylation of FoxO1. In addition, FoxO1 RNAi brought down autophagy induced by high-glucose. Intriguingly, EGCG successfully reversed ROS, autophagy and acetylated FoxO1 in high-glucose treated H9c2 cells. Our findings suggest that FoxO1, especially the acetylated form, regulates high-glucose induced autophagy in H9c2 cardiomyoblasts, which can be prevented by EGCG via a possible ROS-FoxO1 pathway.
Product Description
Supelco
(−)-Epicatechin, analytical standard
Sigma-Aldrich
(−)-Epicatechin, ≥98% (HPLC), from green tea
Sigma-Aldrich
(−)-Epicatechin, ≥90% (HPLC)
Epicatechin, primary reference standard
Supelco
(+)-Catechin, analytical standard
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Epigallocatechin gallate, primary reference standard
Supelco
(−)-Epigallocatechin gallate, analytical standard
Supelco
Epigallocatechin gallate, Pharmaceutical Secondary Standard; Certified Reference Material