Merck
  • Development of a rapid capture-cum-detection method for Escherichia coli O157 from apple juice comprising nano-immunomagnetic separation in tandem with surface enhanced Raman scattering.
Development of a rapid capture-cum-detection method for Escherichia coli O157 from apple juice comprising nano-immunomagnetic separation in tandem with surface enhanced Raman scattering.
International journal of food microbiology (2014-08-19)
Roya Najafi, Shubhasish Mukherjee, Jim Hudson, Anup Sharma, Pratik Banerjee
ABSTRACT
A combined capture and detection method comprising of nano-immunomagnetic separation (NIMS) and surface enhanced Raman spectroscopy (SERS) was developed to detect Escherichia coli O157 from liquid media including apple juice. The capture antibodies (cAbs) were immobilized on magnetite-gold (Fe3O4/Au) magnetic nanoparticles (MNPs) which were used for separation and concentration of the E. coli O157 cells from model liquid food matrix. The capture efficiency (CE) for E. coli O157 using MNP was found to be approximately 84-94%. No cross reactivity was observed with background non-target organisms. There was a significant difference in the mean CE of bacteria captured by MNP and commercially sourced immunomagnetic microbeads (p<0.05). For the detection of target pathogen, SERS labels were prepared by conjugating gold nanoparticles with Raman reporter molecules and the detector antibody (dAb). Au-Raman label-dAb was interacted with gold coated MNP-cAb-E. coli O157 complex. The ability of this immunoassay to detect E. coli O157 in apple juice was investigated. We have successfully applied the synthesized Fe3O4/Au nanoclusters to E. coli O157 detection in apple juice using the SERS method. The lowest detectable bacterial cell concentration in apple juice was 10(2)CFU/mL with a total analysis time of less than an hour. This method presents a convenient way of preconcentration, separation, and detection of low levels of target pathogen from liquid food matrix.
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size, 5 mg/mL in toluene
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 20 nm avg. part. size, 5 mg/mL in toluene
Sigma-Aldrich
Gold, foil, thickness 0.025 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, powder, <850 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Gold, evaporation slug, diam. × L 0.3 cm × 0.6 cm, 99.99% trace metals basis
Sigma-Aldrich
Iron(II,III) oxide, powder, <5 μm, 95%
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Iron(II,III) oxide, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.5 mm, 99.999% trace metals basis
Sigma-Aldrich
Gold coated glass slide, layer thickness 100 Å, 99.999% (Au)
Sigma-Aldrich
Gold coated glass cover slip, 99.999% (Au), diam. × thickness 15 mm × 130-170 μm, layer thickness 100 Å
Sigma-Aldrich
Gold coated glass cover slip, layer thickness 100 Å, 99.999% (Au), L × W × thickness 22 mm × 22 mm × 130-170 μm, square
Sigma-Aldrich
Gold, nanopowder, <100 nm particle size, 99.9% trace metals basis
Sigma-Aldrich
Iron(II,III) oxide, nanopowder, 50-100 nm particle size (SEM), 97% trace metals basis
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size, 5 mg/mL in H2O
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size, 5 mg/mL in H2O
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 20 nm avg. part. size, 5 mg/mL in H2O
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size, 5 mg/mL in toluene
Sigma-Aldrich
Gold coated glass slide, layer thickness 1000 Å, 99.999% (Au)
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size (TEM), PEG functionalized, 1 mg/mL Fe in H2O, dispersion
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size (TEM), carboxylic acid functionalized, 5 mg/mL Fe in H2O, dispersion
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size (TEM), amine functionalized, 1 mg/mL Fe in H2O, dispersion
Gold, insulated wire, 1m, conductor diameter 0.125mm, insulation thickness 0.014mm, polyester insulation, 99.99%
Gold, microfoil, disks, 10mm, thinness 0.75μm, specific density 1449μg/cm2, permanent mylar 3.5μm support, 99.99+%
Gold, insulated wire, 1m, conductor diameter 0.125mm, insulation thickness 0.016mm, PTFE (polytetrafluoroethylene) insulation, 99.99%
Gold, tube, 200mm, outside diameter 1.60mm, inside diameter 0.6mm, wall thickness 0.5mm, as drawn, 99.95%
Gold, rod, 10mm, diameter 5.0mm, as drawn, 99.95%