Merck
  • Differences in relative iron bioavailability in traditional Bangladeshi meal plans.
Differences in relative iron bioavailability in traditional Bangladeshi meal plans.
Food and nutrition bulletin (2015-02-03)
Diane M DellaValle, Raymond P Glahn
ABSTRACT
Iron deficiency is the most common nutrient deficiency worldwide. Large intakes of micronutrient-poor staple crops, coupled with low intakes of highly bioavailable dietary iron, are a major cause of this deficiency. To examine the concentration and relative bioavailability of iron in several models (n = 23) of traditional Bangladeshi meals (rice, lentils/dal, vegetable, and fish), as well as the effect of removal of the lentil seed coat on the nutritional quality of iron. The relative bioavailability of iron was assessed by the in vitro/Caco-2 cell culture method, iron concentration by an inductively coupled argon-plasma emission spectrometer (ICAP-ES), and phytic acid concentration by colorimetric assay. The recipes contained 75% to 85% rice, 0% to 15% dal (containing whole or dehulled lentils), 0% to 15% vegetable curry, and 0% to 8% fish. While the iron concentrations of recipes containing dehulled dal were significantly lower than those of recipes containing whole dal (p = .005), seed coat removal doubled relative iron bioavailability and increased phytic acid concentration (p < .001). The addition of fish to the meals had no significant effect on relative iron bioavailability. Iron concentration and relative iron bioavailability were correlated in the recipes containing dehulled dal (r = 0.48, p = .03), but not whole dal (r = -0.047, p = .84). The total amount of iron absorbed from traditional Bangladeshi meals is dependent upon iron concentration, and dehulling lentils removes inhibitory factors increasing iron uptake but also increases the density of phytic acid in the lentil sample. Thus, along with breeding for high iron concentration and bioavailability (i.e., biofortification), seed coat removal plus measures to lower phytic acid concentrations may be an important strategy to improve the bioavailability of iron in lentils and other pulse crops.
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Iron, IRMM®, certified reference material, 0.5 mm wire
Iron, foil, not light tested, 50x50mm, thickness 0.001mm, 99.85%
Iron, foil, not light tested, 150x150mm, thickness 0.008mm, 99.85%
Iron, foil, 20m coil, thickness 0.075mm, hard, 99.5%
Iron, foil, 15mm disks, thickness 0.38mm, hard, 99.5%
Iron, foil, 10mm disks, thickness 0.075mm, as rolled, 99.99+%
Iron, foil, 15mm disks, thickness 0.008mm, 99.85%
Iron, foil, 10mm disks, thickness 0.075mm, hard, 99.5%
Iron, foil, 25mm disks, thickness 0.003mm, 99.85%
Iron, foil, 1m coil, thickness 0.1mm, hard, 99.5%
Iron, foil, 4mm disks, thickness 0.008mm, 99.85%
Iron, foil, 10mm disks, thickness 0.125mm, as rolled, 99.99+%
Iron, foil, 10mm disks, thickness 0.9mm, as rolled, 99.5%
Iron, foil, 1m coil, thickness 0.075mm, hard, 99.5%
Iron, foil, 15mm disks, thickness 0.05mm, as rolled, 99.99+%
Iron, foil, 15mm disks, thickness 0.038mm, hard, 99.5%
Iron, foil, 1m coil, thickness 0.05mm, hard, 99.5%
Iron, foil, 10mm disks, thickness 1.0mm, as rolled, 99.5%
Iron, foil, 10mm disks, thickness 0.15mm, hard, 99.5%
Iron, foil, 25mm disks, thickness 0.075mm, hard, 99.5%
Iron, foil, 20m coil, thickness 0.038mm, hard, 99.5%
Iron, foil, 4mm disks, thickness 0.003mm, 99.85%
Iron, foil, 1m coil, thickness 0.025mm, hard, 99.5%
Iron, foil, 25mm disks, thickness 0.015mm, 99.85%
Iron, foil, 10mm disks, thickness 0.025mm, hard, 99.5%
Iron, foil, 10mm disks, thickness 0.038mm, hard, 99.5%
Iron, foil, 25mm disks, thickness 1.0mm, as rolled, 99.5%